Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62394381-62394654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIMT1
Entrez Gene ID
27292
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
DIM1, DIMT1L, HSA9761, HUSSY5
UniGene
Hs.731665
Gene Chromosome Location
Chr.5: 62388523-62403939 on Build GRCh38
Transcripts
NM_014473.2 XM_011543323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403677 c.558C>T p.D186D Chr.5: 62394496-62394496 on Build GRCh38
3855805 c.447G>A p.R149R Chr.5: 62394607-62394607 on Build GRCh38
6644021 c.484C>T p.R162* Chr.5: 62394570-62394570 on Build GRCh38
6644022 c.557A>G p.D186G Chr.5: 62394497-62394497 on Build GRCh38
1695814 c.567G>A p.M189I Chr.5: 62394487-62394487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757992850 Chr.5: 62394570-62394570 on Build GRCh38
rs754882649 Chr.5: 62394456-62394456 on Build GRCh38
rs745717280 Chr.5: 62394592-62394592 on Build GRCh38
rs778485250 Chr.5: 62394469-62394469 on Build GRCh38
rs372250999 Chr.5: 62394493-62394493 on Build GRCh38
rs770926646 Chr.5: 62394574-62394574 on Build GRCh38
rs778856297 Chr.5: 62394464-62394464 on Build GRCh38
rs374755893 Chr.5: 62394445-62394446 on Build GRCh38
rs751631127 Chr.5: 62394531-62394531 on Build GRCh38
rs774782434 Chr.5: 62394452-62394453 on Build GRCh38
rs763193253 Chr.5: 62394520-62394520 on Build GRCh38
rs759205859 Chr.5: 62394504-62394504 on Build GRCh38
rs776524614 Chr.5: 62394503-62394503 on Build GRCh38
rs771536766 Chr.5: 62394477-62394477 on Build GRCh38
rs370174707 Chr.5: 62394479-62394479 on Build GRCh38
rs569592415 Chr.5: 62394511-62394511 on Build GRCh38
rs141512628 Chr.5: 62394525-62394525 on Build GRCh38
rs962485722 Chr.5: 62394460-62394463 on Build GRCh38
rs767867155 Chr.5: 62394462-62394467 on Build GRCh38
rs777272669 Chr.5: 62394573-62394573 on Build GRCh38
rs750819640 Chr.5: 62394546-62394546 on Build GRCh38
rs763282145 Chr.5: 62394599-62394599 on Build GRCh38
rs200556696 Chr.5: 62394540-62394540 on Build GRCh38
rs757786036 Chr.5: 62394537-62394537 on Build GRCh38
rs747789972 Chr.5: 62394564-62394564 on Build GRCh38
rs778573002 Chr.5: 62394555-62394555 on Build GRCh38
rs756408984 Chr.5: 62394548-62394548 on Build GRCh38
rs946870178 Chr.5: 62394598-62394598 on Build GRCh38
rs982107903 Chr.5: 62394470-62394470 on Build GRCh38
rs752978750 Chr.5: 62394593-62394594 on Build GRCh38
rs188681034 Chr.5: 62394476-62394476 on Build GRCh38
rs147042946 Chr.5: 62394576-62394576 on Build GRCh38
rs765607181 Chr.5: 62394515-62394515 on Build GRCh38
rs775912795 Chr.5: 62394596-62394596 on Build GRCh38
rs770711458 Chr.5: 62394497-62394497 on Build GRCh38
rs969814325 Chr.5: 62394443-62394443 on Build GRCh38
rs530522421 Chr.5: 62394520-62394523 on Build GRCh38
rs759903414 Chr.5: 62394458-62394459 on Build GRCh38
rs147688707 Chr.5: 62394593-62394593 on Build GRCh38
rs570486274 Chr.5: 62394422-62394422 on Build GRCh38

View Full Product Details