Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74698836-74699075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2I
Entrez Gene ID
2969
Gene Name
general transcription factor IIi
Gene Aliases
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6
UniGene
Hs.647041
Gene Chromosome Location
Chr.7: 74657665-74760692 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

Gene Symbol
LOC101926943
Entrez Gene ID
101926943
Gene Name
uncharacterized LOC101926943
Gene Aliases
-
UniGene
Hs.654772
Gene Chromosome Location
Chr.7: 74688937-74728918 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

SNP Detail

SNP ID Chromosome Location
rs917511936 Chr.7: 74698955-74698955 on Build GRCh38
rs782328272 Chr.7: 74698977-74698977 on Build GRCh38
rs929343959 Chr.7: 74698917-74698917 on Build GRCh38
rs782594544 Chr.7: 74698936-74698936 on Build GRCh38
rs782679698 Chr.7: 74698957-74698957 on Build GRCh38
rs943454473 Chr.7: 74698878-74698878 on Build GRCh38
rs910596522 Chr.7: 74698876-74698876 on Build GRCh38
rs782230656 Chr.7: 74699008-74699008 on Build GRCh38
rs781865877 Chr.7: 74698948-74698948 on Build GRCh38
rs782500588 Chr.7: 74698951-74698951 on Build GRCh38
rs782585870 Chr.7: 74699007-74699007 on Build GRCh38
rs782120808 Chr.7: 74699024-74699024 on Build GRCh38
rs781886172 Chr.7: 74698948-74698948 on Build GRCh38
rs782003787 Chr.7: 74699023-74699023 on Build GRCh38
rs375313162 Chr.7: 74698895-74698895 on Build GRCh38
rs906057032 Chr.7: 74699032-74699032 on Build GRCh38
rs973482232 Chr.7: 74698887-74698887 on Build GRCh38
rs1053566327 Chr.7: 74699044-74699044 on Build GRCh38
rs781973735 Chr.7: 74698947-74698947 on Build GRCh38
rs782345125 Chr.7: 74699013-74699013 on Build GRCh38
rs782514745 Chr.7: 74698948-74698949 on Build GRCh38
rs781931714 Chr.7: 74699040-74699040 on Build GRCh38
rs200063075 Chr.7: 74698963-74698963 on Build GRCh38
rs782413639 Chr.7: 74699035-74699035 on Build GRCh38

View Full Product Details