Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53482879-53483152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K12
Entrez Gene ID
7786
Gene Name
mitogen-activated protein kinase kinase kinase 12
Gene Aliases
DLK, MEKK12, MUK, ZPK, ZPKP1
UniGene
Hs.713539
Gene Chromosome Location
Chr.12: 53480492-53501066 on Build GRCh38
Transcripts
NM_001193511.1 NM_006301.3 XM_005269138.3 XM_006719588.3 XM_011538725.2 XM_017019956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154648 c.1706C>T p.S569L Chr.12: 53483097-53483097 on Build GRCh38
1362712 c.1605delC p.S536fs*45 Chr.12: 53483099-53483099 on Build GRCh38
4575556 c.1687C>T p.R563C Chr.12: 53483017-53483017 on Build GRCh38
5431338 c.1566G>C p.L522L Chr.12: 53483138-53483138 on Build GRCh38
5634876 c.1771C>T p.P591S Chr.12: 53482933-53482933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052023976 Chr.12: 53483131-53483131 on Build GRCh38
rs777368317 Chr.12: 53483099-53483099 on Build GRCh38
rs761114004 Chr.12: 53482949-53482949 on Build GRCh38
rs763110667 Chr.12: 53483045-53483045 on Build GRCh38
rs775911278 Chr.12: 53483042-53483042 on Build GRCh38
rs777812158 Chr.12: 53482907-53482907 on Build GRCh38
rs769564619 Chr.12: 53482927-53482927 on Build GRCh38
rs775823102 Chr.12: 53483104-53483104 on Build GRCh38
rs770855588 Chr.12: 53482912-53482913 on Build GRCh38
rs537027662 Chr.12: 53482976-53482976 on Build GRCh38
rs764385650 Chr.12: 53482972-53482972 on Build GRCh38
rs771720048 Chr.12: 53483020-53483020 on Build GRCh38
rs969396295 Chr.12: 53483033-53483033 on Build GRCh38
rs199578693 Chr.12: 53482921-53482921 on Build GRCh38
rs758006140 Chr.12: 53483092-53483092 on Build GRCh38
rs752739965 Chr.12: 53482901-53482901 on Build GRCh38
rs756170854 Chr.12: 53482988-53482988 on Build GRCh38
rs200925448 Chr.12: 53482938-53482938 on Build GRCh38
rs755903927 Chr.12: 53483073-53483073 on Build GRCh38
rs758665071 Chr.12: 53482904-53482904 on Build GRCh38
rs374436740 Chr.12: 53483061-53483061 on Build GRCh38
rs747818841 Chr.12: 53483019-53483019 on Build GRCh38
rs147211101 Chr.12: 53483088-53483088 on Build GRCh38
rs527895227 Chr.12: 53483057-53483057 on Build GRCh38
rs560592149 Chr.12: 53483017-53483017 on Build GRCh38
rs770090015 Chr.12: 53483103-53483103 on Build GRCh38
rs760073228 Chr.12: 53482958-53482958 on Build GRCh38
rs1050177151 Chr.12: 53483062-53483062 on Build GRCh38
rs370778122 Chr.12: 53483129-53483129 on Build GRCh38
rs779220145 Chr.12: 53483098-53483099 on Build GRCh38
rs780099120 Chr.12: 53483008-53483008 on Build GRCh38
rs138251645 Chr.12: 53482953-53482953 on Build GRCh38
rs756939627 Chr.12: 53482913-53482913 on Build GRCh38
rs746179599 Chr.12: 53483029-53483029 on Build GRCh38
rs761905186 Chr.12: 53483114-53483114 on Build GRCh38
rs777027128 Chr.12: 53483022-53483022 on Build GRCh38
rs144272449 Chr.12: 53483132-53483132 on Build GRCh38
rs769176033 Chr.12: 53483105-53483105 on Build GRCh38
rs143743852 Chr.12: 53483016-53483016 on Build GRCh38
rs765708332 Chr.12: 53482962-53482962 on Build GRCh38
rs750272809 Chr.12: 53483068-53483068 on Build GRCh38
rs200025214 Chr.12: 53482986-53482986 on Build GRCh38
rs367940712 Chr.12: 53483080-53483080 on Build GRCh38
rs959871483 Chr.12: 53482955-53482955 on Build GRCh38
rs967246115 Chr.12: 53483006-53483006 on Build GRCh38
rs764138753 Chr.12: 53483051-53483051 on Build GRCh38
rs533517427 Chr.12: 53482909-53482909 on Build GRCh38
rs898097230 Chr.12: 53483082-53483082 on Build GRCh38
rs890118137 Chr.12: 53482906-53482906 on Build GRCh38
rs777045103 Chr.12: 53482954-53482954 on Build GRCh38
rs549570283 Chr.12: 53483089-53483089 on Build GRCh38
rs150402780 Chr.12: 53482946-53482946 on Build GRCh38
rs757489134 Chr.12: 53482981-53482981 on Build GRCh38
rs368607330 Chr.12: 53482966-53482966 on Build GRCh38
rs770320620 Chr.12: 53483038-53483038 on Build GRCh38
rs754935100 Chr.12: 53483081-53483081 on Build GRCh38
rs745858800 Chr.12: 53482920-53482920 on Build GRCh38
rs766432155 Chr.12: 53483079-53483079 on Build GRCh38
rs781204369 Chr.12: 53482919-53482919 on Build GRCh38
rs960741957 Chr.12: 53482977-53482977 on Build GRCh38
rs925794659 Chr.12: 53483060-53483060 on Build GRCh38
rs750213682 Chr.12: 53482987-53482987 on Build GRCh38
rs749054659 Chr.12: 53482906-53482923 on Build GRCh38
rs775571911 Chr.12: 53482963-53482963 on Build GRCh38

View Full Product Details