Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48136754-48137003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYEF2
Entrez Gene ID
50804
Gene Name
myelin expression factor 2
Gene Aliases
HsT18564, MEF-2, MST156, MSTP156, myEF-2
UniGene
Hs.653123
Gene Chromosome Location
Chr.15: 48134632-48178517 on Build GRCh38
Transcripts
XM_005254425.4 XM_017022285.1 XM_017022286.1 XM_017022287.1 XM_017022291.1 XM_017022292.1 NM_205850.2 XM_017022079.1 XM_017022080.1

Gene Symbol
SLC24A5
Entrez Gene ID
283652
Gene Name
solute carrier family 24 member 5
Gene Aliases
JSX, NCKX5, OCA6, SHEP4
UniGene
Hs.710240
Gene Chromosome Location
Chr.15: 48120972-48142392 on Build GRCh38
Transcripts
XM_005254425.4 XM_017022285.1 XM_017022286.1 XM_017022287.1 XM_017022291.1 XM_017022292.1 NM_205850.2 XM_017022079.1 XM_017022080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780451 c.793G>C p.D265H Chr.15: 48136885-48136885 on Build GRCh38
4559614 c.811G>A p.E271K Chr.15: 48136903-48136903 on Build GRCh38
5888262 c.871G>A p.D291N Chr.15: 48136963-48136963 on Build GRCh38
3706602 c.717G>A p.M239I Chr.15: 48136809-48136809 on Build GRCh38
700389 c.757G>A p.E253K Chr.15: 48136849-48136849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764606204 Chr.15: 48136825-48136825 on Build GRCh38
rs1047344623 Chr.15: 48136873-48136873 on Build GRCh38
rs764396166 Chr.15: 48136930-48136930 on Build GRCh38
rs368088650 Chr.15: 48136824-48136825 on Build GRCh38
rs147513140 Chr.15: 48136921-48136921 on Build GRCh38
rs923066036 Chr.15: 48136868-48136868 on Build GRCh38
rs757960224 Chr.15: 48136965-48136965 on Build GRCh38
rs756606080 Chr.15: 48136824-48136824 on Build GRCh38
rs754321211 Chr.15: 48136941-48136941 on Build GRCh38
rs772412071 Chr.15: 48136911-48136911 on Build GRCh38
rs774586843 Chr.15: 48136789-48136789 on Build GRCh38
rs769822996 Chr.15: 48136887-48136887 on Build GRCh38
rs1028855122 Chr.15: 48136853-48136853 on Build GRCh38
rs926843077 Chr.15: 48136940-48136940 on Build GRCh38
rs886122994 Chr.15: 48136883-48136883 on Build GRCh38
rs150700445 Chr.15: 48136865-48136865 on Build GRCh38
rs575303689 Chr.15: 48136874-48136874 on Build GRCh38
rs796318500 Chr.15: 48136931-48136931 on Build GRCh38
rs759757741 Chr.15: 48136906-48136906 on Build GRCh38
rs556950130 Chr.15: 48136871-48136871 on Build GRCh38
rs936950299 Chr.15: 48136961-48136961 on Build GRCh38
rs138230830 Chr.15: 48136837-48136837 on Build GRCh38
rs769236601 Chr.15: 48136820-48136822 on Build GRCh38
rs761055989 Chr.15: 48136927-48136927 on Build GRCh38
rs140666229 Chr.15: 48136867-48136867 on Build GRCh38
rs149836406 Chr.15: 48136885-48136885 on Build GRCh38
rs369645952 Chr.15: 48136829-48136829 on Build GRCh38
rs141831999 Chr.15: 48136861-48136861 on Build GRCh38
rs753108233 Chr.15: 48136812-48136812 on Build GRCh38
rs745430668 Chr.15: 48136863-48136863 on Build GRCh38
rs146317963 Chr.15: 48136799-48136799 on Build GRCh38
rs758011238 Chr.15: 48136864-48136864 on Build GRCh38
rs376395790 Chr.15: 48136788-48136788 on Build GRCh38
rs756750463 Chr.15: 48136850-48136850 on Build GRCh38
rs562133637 Chr.15: 48136798-48136798 on Build GRCh38
rs989850155 Chr.15: 48136964-48136964 on Build GRCh38
rs751246552 Chr.15: 48136969-48136969 on Build GRCh38
rs774546543 Chr.15: 48136892-48136892 on Build GRCh38
rs779623303 Chr.15: 48136968-48136968 on Build GRCh38
rs373109494 Chr.15: 48136953-48136953 on Build GRCh38
rs768346861 Chr.15: 48136869-48136869 on Build GRCh38
rs765813689 Chr.15: 48136942-48136942 on Build GRCh38
rs754756034 Chr.15: 48136973-48136973 on Build GRCh38

View Full Product Details