Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56136981-56137238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8J3
Entrez Gene ID
81168
Gene Name
olfactory receptor family 8 subfamily J member 3
Gene Aliases
OR11-173
UniGene
Hs.554523
Gene Chromosome Location
Chr.11: 56136771-56137718 on Build GRCh38
Transcripts
NM_001004064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676010 c.595G>C p.V199L Chr.11: 56137124-56137124 on Build GRCh38
4956989 c.673A>G p.I225V Chr.11: 56137046-56137046 on Build GRCh38
688667 c.706A>G p.K236E Chr.11: 56137013-56137013 on Build GRCh38
6992271 c.550C>G p.L184V Chr.11: 56137169-56137169 on Build GRCh38
384884 c.561A>T p.L187F Chr.11: 56137158-56137158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758931012 Chr.11: 56137152-56137152 on Build GRCh38
rs138320062 Chr.11: 56137026-56137026 on Build GRCh38
rs752356052 Chr.11: 56137135-56137135 on Build GRCh38
rs765887369 Chr.11: 56137011-56137011 on Build GRCh38
rs750821999 Chr.11: 56137146-56137146 on Build GRCh38
rs184855718 Chr.11: 56137112-56137112 on Build GRCh38
rs780367713 Chr.11: 56137171-56137171 on Build GRCh38
rs375474573 Chr.11: 56137023-56137023 on Build GRCh38
rs751097194 Chr.11: 56137111-56137111 on Build GRCh38
rs1012113114 Chr.11: 56137027-56137027 on Build GRCh38
rs780178067 Chr.11: 56137059-56137059 on Build GRCh38
rs779306688 Chr.11: 56137142-56137142 on Build GRCh38
rs986768949 Chr.11: 56137039-56137039 on Build GRCh38
rs766290415 Chr.11: 56137107-56137107 on Build GRCh38
rs767975410 Chr.11: 56137136-56137137 on Build GRCh38
rs1044817587 Chr.11: 56137022-56137022 on Build GRCh38
rs930491972 Chr.11: 56137138-56137138 on Build GRCh38
rs768740274 Chr.11: 56137069-56137069 on Build GRCh38
rs779864478 Chr.11: 56137088-56137088 on Build GRCh38
rs772497322 Chr.11: 56137057-56137057 on Build GRCh38
rs755909549 Chr.11: 56137079-56137079 on Build GRCh38
rs747150200 Chr.11: 56137066-56137066 on Build GRCh38
rs1384094 Chr.11: 56137097-56137097 on Build GRCh38
rs767141122 Chr.11: 56137115-56137115 on Build GRCh38
rs140818281 Chr.11: 56137017-56137017 on Build GRCh38
rs746550275 Chr.11: 56137090-56137091 on Build GRCh38
rs758292560 Chr.11: 56137010-56137010 on Build GRCh38
rs779174991 Chr.11: 56137047-56137047 on Build GRCh38
rs112783093 Chr.11: 56137105-56137105 on Build GRCh38
rs750336779 Chr.11: 56137008-56137008 on Build GRCh38
rs1034897541 Chr.11: 56137015-56137015 on Build GRCh38
rs761542650 Chr.11: 56137003-56137003 on Build GRCh38
rs764949781 Chr.11: 56137006-56137006 on Build GRCh38
rs755743354 Chr.11: 56137139-56137139 on Build GRCh38
rs757629356 Chr.11: 56137034-56137034 on Build GRCh38
rs141817251 Chr.11: 56137055-56137055 on Build GRCh38
rs781045638 Chr.11: 56137029-56137029 on Build GRCh38
rs752411061 Chr.11: 56137033-56137033 on Build GRCh38

View Full Product Details