Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 730448-730673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NARFL
Entrez Gene ID
64428
Gene Name
nuclear prelamin A recognition factor like
Gene Aliases
HPRN, IOP1, LET1L, NAR1, PRN
UniGene
Hs.513247
Gene Chromosome Location
Chr.16: 729753-741038 on Build GRCh38
Transcripts
NM_001304799.1 NM_022493.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063010 c.1353G>A p.S451S Chr.16: 730495-730495 on Build GRCh38
6721936 c.1257C>T p.H419H Chr.16: 730591-730591 on Build GRCh38
396957 c.1241G>A p.R414K Chr.16: 730607-730607 on Build GRCh38
6838907 c.1219C>A p.Q407K Chr.16: 730629-730629 on Build GRCh38
4465621 c.1392C>T p.A464A Chr.16: 730456-730456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143166023 Chr.16: 730544-730544 on Build GRCh38
rs757807510 Chr.16: 730543-730543 on Build GRCh38
rs778425484 Chr.16: 730481-730481 on Build GRCh38
rs966669823 Chr.16: 730530-730530 on Build GRCh38
rs759731007 Chr.16: 730521-730521 on Build GRCh38
rs752559860 Chr.16: 730612-730622 on Build GRCh38
rs200260851 Chr.16: 730592-730592 on Build GRCh38
rs748883717 Chr.16: 730552-730552 on Build GRCh38
rs752141515 Chr.16: 730536-730536 on Build GRCh38
rs140239745 Chr.16: 730546-730546 on Build GRCh38
rs749570634 Chr.16: 730626-730626 on Build GRCh38
rs766810490 Chr.16: 730651-730651 on Build GRCh38
rs755192432 Chr.16: 730468-730468 on Build GRCh38
rs750830398 Chr.16: 730583-730583 on Build GRCh38
rs202243971 Chr.16: 730615-730615 on Build GRCh38
rs762468429 Chr.16: 730580-730580 on Build GRCh38
rs763225091 Chr.16: 730574-730574 on Build GRCh38
rs767440685 Chr.16: 730645-730645 on Build GRCh38
rs368018618 Chr.16: 730566-730566 on Build GRCh38
rs772944113 Chr.16: 730565-730565 on Build GRCh38
rs536829915 Chr.16: 730501-730501 on Build GRCh38
rs773032696 Chr.16: 730520-730520 on Build GRCh38
rs752804681 Chr.16: 730473-730473 on Build GRCh38
rs772028390 Chr.16: 730564-730564 on Build GRCh38
rs761989802 Chr.16: 730644-730644 on Build GRCh38
rs35973754 Chr.16: 730603-730603 on Build GRCh38
rs368193380 Chr.16: 730555-730555 on Build GRCh38
rs150311222 Chr.16: 730558-730558 on Build GRCh38
rs574344672 Chr.16: 730590-730590 on Build GRCh38
rs747649200 Chr.16: 730602-730602 on Build GRCh38
rs778446444 Chr.16: 730600-730600 on Build GRCh38
rs774640747 Chr.16: 730641-730641 on Build GRCh38
rs764219549 Chr.16: 730531-730531 on Build GRCh38
rs775803074 Chr.16: 730570-730570 on Build GRCh38
rs374312196 Chr.16: 730551-730551 on Build GRCh38
rs576737498 Chr.16: 730617-730617 on Build GRCh38
rs770570744 Chr.16: 730569-730569 on Build GRCh38
rs137994770 Chr.16: 730575-730575 on Build GRCh38
rs765277097 Chr.16: 730469-730469 on Build GRCh38
rs768089758 Chr.16: 730502-730502 on Build GRCh38
rs144802616 Chr.16: 730545-730545 on Build GRCh38
rs753523131 Chr.16: 730591-730591 on Build GRCh38
rs745546896 Chr.16: 730489-730489 on Build GRCh38
rs373625573 Chr.16: 730482-730482 on Build GRCh38
rs752289658 Chr.16: 730496-730496 on Build GRCh38
rs765488256 Chr.16: 730522-730522 on Build GRCh38
rs767128512 Chr.16: 730507-730508 on Build GRCh38
rs763136041 Chr.16: 730528-730528 on Build GRCh38
rs34851515 Chr.16: 730510-730510 on Build GRCh38
rs142623781 Chr.16: 730495-730495 on Build GRCh38
rs773793404 Chr.16: 730648-730648 on Build GRCh38
rs1048485093 Chr.16: 730618-730618 on Build GRCh38
rs751665967 Chr.16: 730576-730576 on Build GRCh38
rs759038456 Chr.16: 730475-730475 on Build GRCh38
rs752793837 Chr.16: 730526-730526 on Build GRCh38
rs768985274 Chr.16: 730638-730638 on Build GRCh38
rs774288305 Chr.16: 730508-730508 on Build GRCh38
rs776533706 Chr.16: 730623-730623 on Build GRCh38
rs757775383 Chr.16: 730612-730612 on Build GRCh38
rs777777723 Chr.16: 730614-730614 on Build GRCh38
rs768064313 Chr.16: 730582-730582 on Build GRCh38
rs374955438 Chr.16: 730556-730556 on Build GRCh38
rs754031768 Chr.16: 730466-730466 on Build GRCh38
rs7188554 Chr.16: 730517-730517 on Build GRCh38
rs375118802 Chr.16: 730650-730650 on Build GRCh38

View Full Product Details