Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137493348-137493601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLIG3
Entrez Gene ID
167826
Gene Name
oligodendrocyte transcription factor 3
Gene Aliases
Bhlhb7, bHLHe20
UniGene
Hs.195398
Gene Chromosome Location
Chr.6: 137492199-137494394 on Build GRCh38
Transcripts
NM_175747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6172812 c.719C>A p.P240H Chr.6: 137493452-137493452 on Build GRCh38
4618564 c.760G>A p.A254T Chr.6: 137493411-137493411 on Build GRCh38
4625668 c.619G>A p.G207S Chr.6: 137493552-137493552 on Build GRCh38
3781554 c.618C>T p.I206I Chr.6: 137493553-137493553 on Build GRCh38
5799131 c.584C>T p.S195L Chr.6: 137493587-137493587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765168163 Chr.6: 137493431-137493431 on Build GRCh38
rs552686088 Chr.6: 137493464-137493464 on Build GRCh38
rs771495331 Chr.6: 137493484-137493484 on Build GRCh38
rs746219379 Chr.6: 137493502-137493502 on Build GRCh38
rs758248239 Chr.6: 137493444-137493444 on Build GRCh38
rs982494307 Chr.6: 137493516-137493516 on Build GRCh38
rs760762411 Chr.6: 137493515-137493515 on Build GRCh38
rs149762183 Chr.6: 137493523-137493523 on Build GRCh38
rs772628498 Chr.6: 137493506-137493506 on Build GRCh38
rs954689924 Chr.6: 137493381-137493381 on Build GRCh38
rs1002787055 Chr.6: 137493492-137493492 on Build GRCh38
rs190573609 Chr.6: 137493555-137493555 on Build GRCh38
rs893664138 Chr.6: 137493368-137493368 on Build GRCh38
rs768119713 Chr.6: 137493376-137493376 on Build GRCh38
rs774814752 Chr.6: 137493389-137493389 on Build GRCh38
rs370881872 Chr.6: 137493466-137493466 on Build GRCh38
rs200104081 Chr.6: 137493526-137493526 on Build GRCh38
rs146880588 Chr.6: 137493529-137493529 on Build GRCh38
rs779679047 Chr.6: 137493446-137493446 on Build GRCh38
rs752086920 Chr.6: 137493557-137493557 on Build GRCh38
rs775835187 Chr.6: 137493514-137493514 on Build GRCh38
rs764132523 Chr.6: 137493417-137493417 on Build GRCh38
rs1053615786 Chr.6: 137493413-137493413 on Build GRCh38
rs374226571 Chr.6: 137493498-137493498 on Build GRCh38
rs557214414 Chr.6: 137493379-137493379 on Build GRCh38
rs768706576 Chr.6: 137493583-137493583 on Build GRCh38
rs747409902 Chr.6: 137493581-137493581 on Build GRCh38
rs762707298 Chr.6: 137493552-137493552 on Build GRCh38
rs771490973 Chr.6: 137493386-137493386 on Build GRCh38
rs201379739 Chr.6: 137493558-137493558 on Build GRCh38
rs751390423 Chr.6: 137493554-137493554 on Build GRCh38
rs759980127 Chr.6: 137493392-137493392 on Build GRCh38
rs200172504 Chr.6: 137493570-137493570 on Build GRCh38
rs780901552 Chr.6: 137493556-137493556 on Build GRCh38
rs771205548 Chr.6: 137493419-137493420 on Build GRCh38
rs750429448 Chr.6: 137493543-137493543 on Build GRCh38
rs760802172 Chr.6: 137493407-137493407 on Build GRCh38
rs767785444 Chr.6: 137493398-137493398 on Build GRCh38
rs746413898 Chr.6: 137493565-137493565 on Build GRCh38
rs779556533 Chr.6: 137493559-137493559 on Build GRCh38
rs765146024 Chr.6: 137493539-137493539 on Build GRCh38
rs377164598 Chr.6: 137493432-137493432 on Build GRCh38
rs780244755 Chr.6: 137493577-137493577 on Build GRCh38
rs749807038 Chr.6: 137493385-137493385 on Build GRCh38
rs117770313 Chr.6: 137493372-137493372 on Build GRCh38
rs781463188 Chr.6: 137493420-137493422 on Build GRCh38
rs747723910 Chr.6: 137493469-137493469 on Build GRCh38
rs372901879 Chr.6: 137493421-137493421 on Build GRCh38
rs573934871 Chr.6: 137493400-137493400 on Build GRCh38
rs761619619 Chr.6: 137493424-137493424 on Build GRCh38
rs766210194 Chr.6: 137493553-137493553 on Build GRCh38

View Full Product Details