Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46650040-46650307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH1
Entrez Gene ID
23143
Gene Name
leucine rich repeats and calponin homology domain containing 1
Gene Aliases
CHDC1, NP81
UniGene
Hs.507971
Gene Chromosome Location
Chr.13: 46553138-46753041 on Build GRCh38
Transcripts
NM_001164211.1 NM_001164213.1 NM_015116.2 XM_017020483.1 XM_017020484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1677652 c.376A>T p.N126Y Chr.13: 46650269-46650269 on Build GRCh38
4491736 c.385C>G p.H129D Chr.13: 46650278-46650278 on Build GRCh38
6074590 c.346T>A p.L116M Chr.13: 46650239-46650239 on Build GRCh38
6074589 c.346T>A p.L116M Chr.13: 46650239-46650239 on Build GRCh38
4047898 c.367G>A p.E123K Chr.13: 46650260-46650260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145138551 Chr.13: 46650100-46650100 on Build GRCh38
rs369182666 Chr.13: 46650246-46650246 on Build GRCh38
rs902816748 Chr.13: 46650277-46650277 on Build GRCh38
rs113339628 Chr.13: 46650087-46650088 on Build GRCh38
rs914307507 Chr.13: 46650229-46650229 on Build GRCh38
rs779511236 Chr.13: 46650223-46650224 on Build GRCh38
rs148383064 Chr.13: 46650101-46650101 on Build GRCh38
rs763394900 Chr.13: 46650184-46650184 on Build GRCh38
rs796625257 Chr.13: 46650100-46650101 on Build GRCh38
rs752533558 Chr.13: 46650270-46650270 on Build GRCh38
rs774785430 Chr.13: 46650128-46650128 on Build GRCh38
rs776666870 Chr.13: 46650158-46650158 on Build GRCh38
rs373174083 Chr.13: 46650253-46650253 on Build GRCh38
rs754902554 Chr.13: 46650262-46650262 on Build GRCh38
rs777701442 Chr.13: 46650280-46650280 on Build GRCh38
rs749949643 Chr.13: 46650239-46650239 on Build GRCh38
rs894283241 Chr.13: 46650078-46650079 on Build GRCh38
rs368757880 Chr.13: 46650170-46650170 on Build GRCh38
rs764515979 Chr.13: 46650193-46650193 on Build GRCh38
rs375002309 Chr.13: 46650220-46650220 on Build GRCh38
rs757452926 Chr.13: 46650272-46650272 on Build GRCh38
rs150516332 Chr.13: 46650160-46650160 on Build GRCh38
rs775995559 Chr.13: 46650182-46650182 on Build GRCh38
rs765818499 Chr.13: 46650248-46650248 on Build GRCh38
rs894439750 Chr.13: 46650084-46650084 on Build GRCh38
rs1023660231 Chr.13: 46650089-46650089 on Build GRCh38
rs767337573 Chr.13: 46650233-46650233 on Build GRCh38
rs912741723 Chr.13: 46650186-46650186 on Build GRCh38
rs903263297 Chr.13: 46650126-46650126 on Build GRCh38
rs182473321 Chr.13: 46650148-46650148 on Build GRCh38
rs996744866 Chr.13: 46650198-46650198 on Build GRCh38
rs778646694 Chr.13: 46650263-46650263 on Build GRCh38
rs774900445 Chr.13: 46650214-46650214 on Build GRCh38
rs111362992 Chr.13: 46650156-46650156 on Build GRCh38

View Full Product Details