Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 61683479-61683714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM2D1
Entrez Gene ID
83941
Gene Name
TM2 domain containing 1
Gene Aliases
BBP
UniGene
Hs.656790
Gene Chromosome Location
Chr.1: 61681047-61725423 on Build GRCh38
Transcripts
NM_032027.2 NR_135160.1 NR_135161.1 NR_135162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398639 c.736G>T p.D246Y Chr.1: 61683510-61683510 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs925598642 Chr.1: 61683664-61683664 on Build GRCh38
rs936913387 Chr.1: 61683676-61683676 on Build GRCh38
rs769747621 Chr.1: 61683516-61683516 on Build GRCh38
rs745833382 Chr.1: 61683505-61683505 on Build GRCh38
rs752820155 Chr.1: 61683685-61683685 on Build GRCh38
rs992162333 Chr.1: 61683508-61683508 on Build GRCh38
rs762730015 Chr.1: 61683557-61683557 on Build GRCh38
rs12134205 Chr.1: 61683565-61683565 on Build GRCh38
rs985494185 Chr.1: 61683688-61683688 on Build GRCh38
rs1021808786 Chr.1: 61683633-61683633 on Build GRCh38
rs1052628087 Chr.1: 61683687-61683687 on Build GRCh38
rs775322312 Chr.1: 61683546-61683546 on Build GRCh38
rs781138548 Chr.1: 61683602-61683605 on Build GRCh38
rs969330674 Chr.1: 61683563-61683563 on Build GRCh38
rs61498236 Chr.1: 61683683-61683683 on Build GRCh38
rs774567500 Chr.1: 61683580-61683580 on Build GRCh38

View Full Product Details