Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 129845662-129845923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM244
Entrez Gene ID
253582
Gene Name
transmembrane protein 244
Gene Aliases
C6orf191, bA174C7.4
UniGene
Hs.448372
Gene Chromosome Location
Chr.6: 129831244-129861622 on Build GRCh38
Transcripts
NM_001010876.1 XM_011535669.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159644 c.118G>C p.E40Q Chr.6: 129845768-129845768 on Build GRCh38
50557 c.109G>A p.V37M Chr.6: 129845777-129845777 on Build GRCh38
4406980 c.119+4A>C p.? Chr.6: 129845763-129845763 on Build GRCh38
5869276 c.92C>T p.S31F Chr.6: 129845794-129845794 on Build GRCh38
1073174 c.67C>A p.L23I Chr.6: 129845819-129845819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146932263 Chr.6: 129845854-129845854 on Build GRCh38
rs953704746 Chr.6: 129845796-129845796 on Build GRCh38
rs17396809 Chr.6: 129845763-129845763 on Build GRCh38
rs375499591 Chr.6: 129845884-129845884 on Build GRCh38
rs961016291 Chr.6: 129845821-129845821 on Build GRCh38
rs143949103 Chr.6: 129845777-129845777 on Build GRCh38
rs1016462662 Chr.6: 129845839-129845839 on Build GRCh38
rs9321192 Chr.6: 129845881-129845881 on Build GRCh38
rs773464896 Chr.6: 129845772-129845772 on Build GRCh38
rs756942968 Chr.6: 129845806-129845806 on Build GRCh38
rs369901389 Chr.6: 129845786-129845786 on Build GRCh38
rs17475221 Chr.6: 129845726-129845726 on Build GRCh38
rs773184767 Chr.6: 129845778-129845778 on Build GRCh38
rs761906646 Chr.6: 129845762-129845762 on Build GRCh38
rs777559831 Chr.6: 129845794-129845794 on Build GRCh38
rs376205331 Chr.6: 129845724-129845724 on Build GRCh38
rs769214869 Chr.6: 129845754-129845754 on Build GRCh38
rs777616692 Chr.6: 129845721-129845721 on Build GRCh38
rs754830761 Chr.6: 129845783-129845783 on Build GRCh38
rs769930408 Chr.6: 129845889-129845889 on Build GRCh38
rs913895202 Chr.6: 129845704-129845704 on Build GRCh38
rs768999035 Chr.6: 129845877-129845877 on Build GRCh38
rs749003577 Chr.6: 129845795-129845795 on Build GRCh38
rs755776898 Chr.6: 129845718-129845718 on Build GRCh38
rs747897499 Chr.6: 129845758-129845758 on Build GRCh38
rs945407364 Chr.6: 129845713-129845713 on Build GRCh38
rs561944279 Chr.6: 129845697-129845697 on Build GRCh38
rs529229542 Chr.6: 129845716-129845716 on Build GRCh38
rs900906701 Chr.6: 129845773-129845773 on Build GRCh38
rs778886450 Chr.6: 129845787-129845787 on Build GRCh38
rs746201105 Chr.6: 129845722-129845722 on Build GRCh38
rs747358015 Chr.6: 129845831-129845831 on Build GRCh38
rs145670153 Chr.6: 129845774-129845774 on Build GRCh38
rs780212414 Chr.6: 129845841-129845841 on Build GRCh38
rs367909541 Chr.6: 129845785-129845785 on Build GRCh38
rs755723669 Chr.6: 129845792-129845792 on Build GRCh38

View Full Product Details