Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 76867559-76867830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GALNAC5
Entrez Gene ID
81849
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
SIAT7-E, SIAT7E, ST6GalNAcV
UniGene
Hs.149201
Gene Chromosome Location
Chr.1: 76867408-77069167 on Build GRCh38
Transcripts
NM_001320273.1 NM_001320274.1 NM_030965.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6231525 c.10C>G p.L4V Chr.1: 76867685-76867685 on Build GRCh38
4915292 c.6G>A p.K2K Chr.1: 76867681-76867681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757664002 Chr.1: 76867694-76867694 on Build GRCh38
rs555015247 Chr.1: 76867772-76867772 on Build GRCh38
rs750613765 Chr.1: 76867699-76867699 on Build GRCh38
rs764756456 Chr.1: 76867665-76867665 on Build GRCh38
rs746771336 Chr.1: 76867719-76867719 on Build GRCh38
rs930329177 Chr.1: 76867799-76867799 on Build GRCh38
rs978113781 Chr.1: 76867582-76867582 on Build GRCh38
rs924001526 Chr.1: 76867595-76867595 on Build GRCh38
rs919134478 Chr.1: 76867777-76867777 on Build GRCh38
rs1054280358 Chr.1: 76867636-76867636 on Build GRCh38
rs999512361 Chr.1: 76867583-76867583 on Build GRCh38
rs751911412 Chr.1: 76867683-76867683 on Build GRCh38
rs776313305 Chr.1: 76867726-76867726 on Build GRCh38
rs759142360 Chr.1: 76867653-76867653 on Build GRCh38
rs772147783 Chr.1: 76867629-76867629 on Build GRCh38
rs936776568 Chr.1: 76867632-76867632 on Build GRCh38
rs113659117 Chr.1: 76867616-76867616 on Build GRCh38
rs1040930670 Chr.1: 76867578-76867578 on Build GRCh38
rs781237013 Chr.1: 76867696-76867696 on Build GRCh38
rs752166312 Chr.1: 76867667-76867667 on Build GRCh38
rs576506278 Chr.1: 76867747-76867747 on Build GRCh38
rs954333872 Chr.1: 76867761-76867761 on Build GRCh38
rs558164719 Chr.1: 76867746-76867746 on Build GRCh38
rs572703660 Chr.1: 76867682-76867682 on Build GRCh38
rs113208751 Chr.1: 76867640-76867640 on Build GRCh38
rs76305167 Chr.1: 76867670-76867670 on Build GRCh38
rs910332243 Chr.1: 76867806-76867806 on Build GRCh38
rs377131012 Chr.1: 76867615-76867615 on Build GRCh38
rs1011546473 Chr.1: 76867661-76867661 on Build GRCh38
rs541709454 Chr.1: 76867794-76867794 on Build GRCh38
rs543948201 Chr.1: 76867752-76867752 on Build GRCh38
rs369888538 Chr.1: 76867710-76867710 on Build GRCh38
rs1002476869 Chr.1: 76867688-76867688 on Build GRCh38
rs995184881 Chr.1: 76867739-76867739 on Build GRCh38
rs780122805 Chr.1: 76867712-76867712 on Build GRCh38
rs770722297 Chr.1: 76867723-76867723 on Build GRCh38
rs569520345 Chr.1: 76867599-76867599 on Build GRCh38
rs115959772 Chr.1: 76867716-76867716 on Build GRCh38
rs972398813 Chr.1: 76867762-76867762 on Build GRCh38
rs770468707 Chr.1: 76867642-76867642 on Build GRCh38
rs749382641 Chr.1: 76867714-76867714 on Build GRCh38
rs772129151 Chr.1: 76867715-76867715 on Build GRCh38
rs948582761 Chr.1: 76867641-76867641 on Build GRCh38
rs773074597 Chr.1: 76867633-76867633 on Build GRCh38
rs562332737 Chr.1: 76867758-76867758 on Build GRCh38
rs1031708945 Chr.1: 76867588-76867588 on Build GRCh38
rs1048682561 Chr.1: 76867804-76867804 on Build GRCh38
rs776387581 Chr.1: 76867647-76867647 on Build GRCh38

View Full Product Details