Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108382292-108382563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT1C4
Entrez Gene ID
27233
Gene Name
sulfotransferase family 1C member 4
Gene Aliases
SULT1C, SULT1C2
UniGene
Hs.312644
Gene Chromosome Location
Chr.2: 108377911-108388989 on Build GRCh38
Transcripts
NM_001321770.1 NM_006588.3 NR_135776.1 NR_135779.1 XM_017003807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5640702 c.333C>T p.I111I Chr.2: 108382422-108382422 on Build GRCh38
5525644 c.336G>A p.L112L Chr.2: 108382425-108382425 on Build GRCh38
1326010 c.334C>A p.L112M Chr.2: 108382423-108382423 on Build GRCh38
6247046 c.301G>T p.E101* Chr.2: 108382390-108382390 on Build GRCh38
1582479 c.391A>G p.K131E Chr.2: 108382480-108382480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770544653 Chr.2: 108382355-108382355 on Build GRCh38
rs374787657 Chr.2: 108382368-108382368 on Build GRCh38
rs548847736 Chr.2: 108382345-108382345 on Build GRCh38
rs374220220 Chr.2: 108382508-108382508 on Build GRCh38
rs372512725 Chr.2: 108382367-108382367 on Build GRCh38
rs762309040 Chr.2: 108382375-108382375 on Build GRCh38
rs949770113 Chr.2: 108382400-108382400 on Build GRCh38
rs41428444 Chr.2: 108382324-108382324 on Build GRCh38
rs41503048 Chr.2: 108382343-108382343 on Build GRCh38
rs374478497 Chr.2: 108382342-108382343 on Build GRCh38
rs745330772 Chr.2: 108382480-108382480 on Build GRCh38
rs537771943 Chr.2: 108382435-108382435 on Build GRCh38
rs760801583 Chr.2: 108382417-108382417 on Build GRCh38
rs754371305 Chr.2: 108382342-108382342 on Build GRCh38
rs780780980 Chr.2: 108382477-108382477 on Build GRCh38
rs996811232 Chr.2: 108382404-108382404 on Build GRCh38
rs746303887 Chr.2: 108382366-108382366 on Build GRCh38
rs369096265 Chr.2: 108382441-108382441 on Build GRCh38
rs749819824 Chr.2: 108382359-108382359 on Build GRCh38
rs142885329 Chr.2: 108382418-108382418 on Build GRCh38
rs200909127 Chr.2: 108382338-108382339 on Build GRCh38
rs765115421 Chr.2: 108382432-108382432 on Build GRCh38
rs201855352 Chr.2: 108382470-108382470 on Build GRCh38
rs772630484 Chr.2: 108382510-108382510 on Build GRCh38
rs770836069 Chr.2: 108382327-108382327 on Build GRCh38
rs776591221 Chr.2: 108382498-108382498 on Build GRCh38
rs752596847 Chr.2: 108382433-108382433 on Build GRCh38
rs376204799 Chr.2: 108382523-108382523 on Build GRCh38
rs777541971 Chr.2: 108382446-108382446 on Build GRCh38
rs767973805 Chr.2: 108382390-108382390 on Build GRCh38
rs768994968 Chr.2: 108382366-108382366 on Build GRCh38
rs34003233 Chr.2: 108382342-108382342 on Build GRCh38
rs1015656617 Chr.2: 108382526-108382526 on Build GRCh38
rs758705139 Chr.2: 108382369-108382369 on Build GRCh38
rs758074758 Chr.2: 108382343-108382343 on Build GRCh38
rs746672220 Chr.2: 108382464-108382464 on Build GRCh38
rs145051924 Chr.2: 108382341-108382342 on Build GRCh38
rs754961720 Chr.2: 108382431-108382431 on Build GRCh38
rs200696314 Chr.2: 108382425-108382425 on Build GRCh38
rs978700668 Chr.2: 108382331-108382331 on Build GRCh38
rs961482462 Chr.2: 108382528-108382528 on Build GRCh38
rs773624943 Chr.2: 108382410-108382410 on Build GRCh38
rs769317251 Chr.2: 108382481-108382481 on Build GRCh38
rs775049384 Chr.2: 108382484-108382484 on Build GRCh38
rs746653855 Chr.2: 108382354-108382354 on Build GRCh38

View Full Product Details