Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 93574631-93574876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS2
Entrez Gene ID
8835
Gene Name
suppressor of cytokine signaling 2
Gene Aliases
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2
UniGene
Hs.485572
Gene Chromosome Location
Chr.12: 93569822-93626236 on Build GRCh38
Transcripts
NM_001270467.1 NM_001270468.1 NM_001270469.1 NM_001270470.1 NM_001270471.1 NM_003877.4 XM_011538929.1 XM_011538935.1 XM_011538936.1 XM_017020147.1 XM_017020148.1 XM_017020149.1 XM_017020150.1 XM_017020151.1 XM_017020152.1 XM_017020153.1 XM_017020154.1 XM_017020155.1 XM_017020156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4728884 c.202G>T p.G68* Chr.12: 93574784-93574784 on Build GRCh38
116078 c.215T>A p.I72N Chr.12: 93574797-93574797 on Build GRCh38
1582843 c.286C>T p.R96* Chr.12: 93574868-93574868 on Build GRCh38
4728885 c.228G>A p.S76S Chr.12: 93574810-93574810 on Build GRCh38
6786707 c.203G>T p.G68V Chr.12: 93574785-93574785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763643934 Chr.12: 93574743-93574743 on Build GRCh38
rs11107113 Chr.12: 93574663-93574663 on Build GRCh38
rs3741676 Chr.12: 93574737-93574737 on Build GRCh38
rs770597376 Chr.12: 93574700-93574700 on Build GRCh38
rs745665725 Chr.12: 93574719-93574719 on Build GRCh38
rs758691020 Chr.12: 93574810-93574810 on Build GRCh38
rs372716188 Chr.12: 93574687-93574687 on Build GRCh38
rs751883912 Chr.12: 93574823-93574823 on Build GRCh38
rs148346233 Chr.12: 93574809-93574809 on Build GRCh38
rs762344209 Chr.12: 93574795-93574795 on Build GRCh38
rs774154101 Chr.12: 93574716-93574716 on Build GRCh38
rs760139379 Chr.12: 93574711-93574711 on Build GRCh38
rs201377221 Chr.12: 93574722-93574722 on Build GRCh38
rs766700768 Chr.12: 93574811-93574811 on Build GRCh38
rs777835642 Chr.12: 93574674-93574674 on Build GRCh38
rs755085956 Chr.12: 93574835-93574835 on Build GRCh38
rs571712132 Chr.12: 93574762-93574762 on Build GRCh38
rs755175668 Chr.12: 93574671-93574671 on Build GRCh38
rs771691402 Chr.12: 93574735-93574735 on Build GRCh38

View Full Product Details