Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26187082-26187331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
NM_001168241.1 NM_001191033.1 XM_006711951.3 XM_011532564.2 XM_011532565.2 XM_011532566.2 XM_011532567.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344751 c.1603G>T p.G535C Chr.2: 26187235-26187235 on Build GRCh38
1738717 c.1634C>T p.P545L Chr.2: 26187266-26187266 on Build GRCh38
5396723 c.1647C>T p.S549S Chr.2: 26187279-26187279 on Build GRCh38
3962703 c.1599-2A>G p.? Chr.2: 26187229-26187229 on Build GRCh38
4751513 c.1679G>T p.C560F Chr.2: 26187311-26187311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944972224 Chr.2: 26187221-26187221 on Build GRCh38
rs1046627949 Chr.2: 26187313-26187313 on Build GRCh38
rs749003017 Chr.2: 26187295-26187295 on Build GRCh38
rs755991297 Chr.2: 26187273-26187273 on Build GRCh38
rs936677530 Chr.2: 26187263-26187263 on Build GRCh38
rs200147579 Chr.2: 26187241-26187241 on Build GRCh38
rs757284825 Chr.2: 26187123-26187123 on Build GRCh38
rs949569791 Chr.2: 26187304-26187304 on Build GRCh38
rs777814755 Chr.2: 26187276-26187276 on Build GRCh38
rs1006108401 Chr.2: 26187107-26187107 on Build GRCh38
rs180805443 Chr.2: 26187197-26187197 on Build GRCh38
rs930292772 Chr.2: 26187271-26187271 on Build GRCh38
rs375192939 Chr.2: 26187264-26187264 on Build GRCh38
rs751209360 Chr.2: 26187211-26187211 on Build GRCh38
rs978166952 Chr.2: 26187234-26187234 on Build GRCh38
rs964573127 Chr.2: 26187134-26187134 on Build GRCh38
rs991046346 Chr.2: 26187268-26187268 on Build GRCh38
rs780782107 Chr.2: 26187261-26187261 on Build GRCh38
rs190900512 Chr.2: 26187135-26187135 on Build GRCh38
rs527902113 Chr.2: 26187207-26187207 on Build GRCh38
rs986692965 Chr.2: 26187170-26187170 on Build GRCh38
rs533731185 Chr.2: 26187176-26187176 on Build GRCh38
rs544207811 Chr.2: 26187116-26187116 on Build GRCh38
rs770718279 Chr.2: 26187301-26187301 on Build GRCh38
rs776677735 Chr.2: 26187099-26187100 on Build GRCh38
rs975923125 Chr.2: 26187175-26187175 on Build GRCh38
rs140565948 Chr.2: 26187189-26187189 on Build GRCh38
rs912037190 Chr.2: 26187212-26187212 on Build GRCh38
rs369234791 Chr.2: 26187266-26187266 on Build GRCh38
rs375018507 Chr.2: 26187265-26187265 on Build GRCh38
rs750736010 Chr.2: 26187182-26187182 on Build GRCh38
rs751296978 Chr.2: 26187121-26187121 on Build GRCh38

View Full Product Details