Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17294065-17294338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
UniGene
Hs.515664
Gene Chromosome Location
Chr.19: 17292131-17303473 on Build GRCh38
Transcripts
NM_024527.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013399 c.1120C>T p.P374S Chr.19: 17294317-17294317 on Build GRCh38
4616235 c.1146C>G p.A382A Chr.19: 17294291-17294291 on Build GRCh38
564910 c.1147G>T p.E383* Chr.19: 17294290-17294290 on Build GRCh38
5013398 c.1147G>A p.E383K Chr.19: 17294290-17294290 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs998316977 Chr.19: 17294317-17294317 on Build GRCh38
rs759189290 Chr.19: 17294157-17294157 on Build GRCh38
rs147460934 Chr.19: 17294279-17294279 on Build GRCh38
rs1039561725 Chr.19: 17294299-17294299 on Build GRCh38
rs747519356 Chr.19: 17294160-17294160 on Build GRCh38
rs1053616627 Chr.19: 17294110-17294110 on Build GRCh38
rs977652491 Chr.19: 17294161-17294161 on Build GRCh38
rs749967311 Chr.19: 17294266-17294266 on Build GRCh38
rs534779612 Chr.19: 17294216-17294216 on Build GRCh38
rs1040202977 Chr.19: 17294156-17294156 on Build GRCh38
rs982175884 Chr.19: 17294211-17294211 on Build GRCh38
rs141047671 Chr.19: 17294159-17294159 on Build GRCh38
rs1005800092 Chr.19: 17294177-17294177 on Build GRCh38
rs1006944370 Chr.19: 17294137-17294137 on Build GRCh38
rs751508698 Chr.19: 17294247-17294247 on Build GRCh38
rs964654470 Chr.19: 17294113-17294113 on Build GRCh38
rs1009034203 Chr.19: 17294236-17294236 on Build GRCh38
rs199521781 Chr.19: 17294316-17294316 on Build GRCh38
rs915046679 Chr.19: 17294277-17294277 on Build GRCh38
rs373430976 Chr.19: 17294280-17294280 on Build GRCh38
rs781444104 Chr.19: 17294209-17294209 on Build GRCh38
rs539922088 Chr.19: 17294186-17294186 on Build GRCh38
rs570862021 Chr.19: 17294180-17294180 on Build GRCh38
rs755368498 Chr.19: 17294295-17294295 on Build GRCh38
rs575619651 Chr.19: 17294188-17294188 on Build GRCh38
rs565854431 Chr.19: 17294214-17294214 on Build GRCh38
rs763851970 Chr.19: 17294243-17294243 on Build GRCh38
rs915846860 Chr.19: 17294184-17294184 on Build GRCh38
rs1010880548 Chr.19: 17294138-17294138 on Build GRCh38
rs749002268 Chr.19: 17294290-17294290 on Build GRCh38
rs1039238148 Chr.19: 17294240-17294240 on Build GRCh38
rs7507125 Chr.19: 17294233-17294233 on Build GRCh38
rs1025674607 Chr.19: 17294164-17294164 on Build GRCh38
rs530828163 Chr.19: 17294150-17294150 on Build GRCh38
rs948329004 Chr.19: 17294185-17294186 on Build GRCh38
rs375212540 Chr.19: 17294273-17294273 on Build GRCh38
rs368930959 Chr.19: 17294176-17294176 on Build GRCh38
rs143298280 Chr.19: 17294257-17294257 on Build GRCh38
rs893646527 Chr.19: 17294136-17294136 on Build GRCh38
rs6512183 Chr.19: 17294201-17294201 on Build GRCh38
rs935195782 Chr.19: 17294232-17294232 on Build GRCh38
rs377027197 Chr.19: 17294282-17294282 on Build GRCh38
rs1053709500 Chr.19: 17294245-17294245 on Build GRCh38
rs757156572 Chr.19: 17294248-17294248 on Build GRCh38
rs890653118 Chr.19: 17294234-17294234 on Build GRCh38

View Full Product Details