Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56474681-56474944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLS2
Entrez Gene ID
27165
Gene Name
glutaminase 2
Gene Aliases
GA, GLS, LGA, hLGA
UniGene
Hs.212606
Gene Chromosome Location
Chr.12: 56470944-56488414 on Build GRCh38
Transcripts
NM_001280796.1 NM_001280797.1 NM_001280798.1 NM_013267.3 XM_005268797.1 XM_017019180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835942 c.1038C>G p.L346L Chr.12: 56474855-56474855 on Build GRCh38
7189262 c.1077A>T p.S359S Chr.12: 56474691-56474691 on Build GRCh38
4687248 c.1032T>C p.L344L Chr.12: 56474861-56474861 on Build GRCh38
1637997 c.1053T>C p.C351C Chr.12: 56474715-56474715 on Build GRCh38
4043638 c.1016A>G p.D339G Chr.12: 56474877-56474877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759926117 Chr.12: 56474910-56474910 on Build GRCh38
rs771446245 Chr.12: 56474899-56474899 on Build GRCh38
rs61731958 Chr.12: 56474882-56474882 on Build GRCh38
rs765467972 Chr.12: 56474768-56474768 on Build GRCh38
rs781036223 Chr.12: 56474703-56474703 on Build GRCh38
rs372835857 Chr.12: 56474806-56474806 on Build GRCh38
rs766646643 Chr.12: 56474839-56474841 on Build GRCh38
rs752878614 Chr.12: 56474795-56474795 on Build GRCh38
rs758485801 Chr.12: 56474796-56474796 on Build GRCh38
rs759726611 Chr.12: 56474765-56474765 on Build GRCh38
rs745543047 Chr.12: 56474714-56474714 on Build GRCh38
rs931761366 Chr.12: 56474788-56474788 on Build GRCh38
rs755309973 Chr.12: 56474865-56474866 on Build GRCh38
rs755709398 Chr.12: 56474786-56474787 on Build GRCh38
rs369037405 Chr.12: 56474812-56474812 on Build GRCh38
rs369561441 Chr.12: 56474734-56474734 on Build GRCh38
rs923600307 Chr.12: 56474711-56474711 on Build GRCh38
rs768341870 Chr.12: 56474879-56474879 on Build GRCh38
rs765526501 Chr.12: 56474914-56474914 on Build GRCh38
rs767036919 Chr.12: 56474731-56474732 on Build GRCh38
rs779853749 Chr.12: 56474867-56474867 on Build GRCh38
rs747514729 Chr.12: 56474889-56474889 on Build GRCh38
rs118135129 Chr.12: 56474752-56474752 on Build GRCh38
rs147026240 Chr.12: 56474923-56474923 on Build GRCh38
rs200412931 Chr.12: 56474728-56474728 on Build GRCh38
rs780767148 Chr.12: 56474716-56474716 on Build GRCh38
rs761234824 Chr.12: 56474742-56474742 on Build GRCh38
rs777024448 Chr.12: 56474753-56474753 on Build GRCh38
rs755930387 Chr.12: 56474866-56474866 on Build GRCh38
rs188729267 Chr.12: 56474775-56474775 on Build GRCh38
rs894671458 Chr.12: 56474857-56474857 on Build GRCh38
rs368844270 Chr.12: 56474776-56474776 on Build GRCh38
rs777335978 Chr.12: 56474866-56474867 on Build GRCh38
rs562890360 Chr.12: 56474823-56474823 on Build GRCh38
rs372992124 Chr.12: 56474822-56474822 on Build GRCh38
rs576524440 Chr.12: 56474846-56474846 on Build GRCh38
rs775231560 Chr.12: 56474725-56474725 on Build GRCh38
rs551221770 Chr.12: 56474903-56474903 on Build GRCh38
rs776954313 Chr.12: 56474905-56474905 on Build GRCh38
rs769545052 Chr.12: 56474723-56474723 on Build GRCh38
rs748894844 Chr.12: 56474871-56474871 on Build GRCh38
rs752017742 Chr.12: 56474864-56474865 on Build GRCh38
rs375271003 Chr.12: 56474921-56474921 on Build GRCh38
rs750942700 Chr.12: 56474727-56474730 on Build GRCh38
rs377211150 Chr.12: 56474808-56474808 on Build GRCh38
rs751517700 Chr.12: 56474810-56474810 on Build GRCh38
rs117078178 Chr.12: 56474804-56474804 on Build GRCh38
rs753517065 Chr.12: 56474869-56474870 on Build GRCh38
rs1036694507 Chr.12: 56474855-56474855 on Build GRCh38

View Full Product Details