Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56645732-56645999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
UniGene
Hs.406510
Gene Chromosome Location
Chr.12: 56638175-56646068 on Build GRCh38
Transcripts
NM_001686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5821746 c.42C>T p.S14S Chr.12: 56645922-56645922 on Build GRCh38
7097834 c.27C>A p.A9A Chr.12: 56645937-56645937 on Build GRCh38
135597 c.121C>T p.H41Y Chr.12: 56645843-56645843 on Build GRCh38
3739786 c.85G>T p.A29S Chr.12: 56645879-56645879 on Build GRCh38
6045778 c.127+1G>A p.? Chr.12: 56645836-56645836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774183285 Chr.12: 56645824-56645824 on Build GRCh38
rs756491836 Chr.12: 56645871-56645871 on Build GRCh38
rs745591336 Chr.12: 56645973-56645973 on Build GRCh38
rs552552913 Chr.12: 56645771-56645771 on Build GRCh38
rs367651199 Chr.12: 56645845-56645845 on Build GRCh38
rs374939855 Chr.12: 56645944-56645944 on Build GRCh38
rs750807220 Chr.12: 56645865-56645865 on Build GRCh38
rs774950891 Chr.12: 56645981-56645981 on Build GRCh38
rs772954458 Chr.12: 56645892-56645892 on Build GRCh38
rs777798405 Chr.12: 56645965-56645965 on Build GRCh38
rs915048004 Chr.12: 56645879-56645879 on Build GRCh38
rs1034147387 Chr.12: 56645975-56645975 on Build GRCh38
rs144845797 Chr.12: 56645891-56645891 on Build GRCh38
rs758351439 Chr.12: 56645951-56645951 on Build GRCh38
rs563393027 Chr.12: 56645933-56645933 on Build GRCh38
rs778926995 Chr.12: 56645942-56645942 on Build GRCh38
rs746919715 Chr.12: 56645966-56645966 on Build GRCh38
rs767663196 Chr.12: 56645777-56645777 on Build GRCh38
rs748523188 Chr.12: 56645776-56645776 on Build GRCh38
rs746819199 Chr.12: 56645893-56645893 on Build GRCh38
rs369108143 Chr.12: 56645970-56645970 on Build GRCh38
rs778276673 Chr.12: 56645787-56645787 on Build GRCh38
rs149422069 Chr.12: 56645847-56645847 on Build GRCh38
rs559141208 Chr.12: 56645773-56645773 on Build GRCh38
rs775064847 Chr.12: 56645921-56645921 on Build GRCh38
rs752067018 Chr.12: 56645794-56645794 on Build GRCh38
rs757773553 Chr.12: 56645855-56645855 on Build GRCh38
rs760309857 Chr.12: 56645846-56645846 on Build GRCh38
rs780442752 Chr.12: 56645872-56645872 on Build GRCh38
rs1023882755 Chr.12: 56645759-56645759 on Build GRCh38
rs200568028 Chr.12: 56645929-56645929 on Build GRCh38
rs764712544 Chr.12: 56645917-56645917 on Build GRCh38
rs770762349 Chr.12: 56645968-56645968 on Build GRCh38
rs2293450 Chr.12: 56645890-56645890 on Build GRCh38
rs771811291 Chr.12: 56645840-56645840 on Build GRCh38
rs746139519 Chr.12: 56645800-56645800 on Build GRCh38
rs781598261 Chr.12: 56645798-56645798 on Build GRCh38
rs750848875 Chr.12: 56645936-56645936 on Build GRCh38
rs961983207 Chr.12: 56645822-56645822 on Build GRCh38
rs865941423 Chr.12: 56645946-56645946 on Build GRCh38
rs759165869 Chr.12: 56645902-56645902 on Build GRCh38
rs368524880 Chr.12: 56645853-56645853 on Build GRCh38
rs755237119 Chr.12: 56645941-56645941 on Build GRCh38
rs754158869 Chr.12: 56645938-56645938 on Build GRCh38
rs770140331 Chr.12: 56645807-56645807 on Build GRCh38
rs371408519 Chr.12: 56645937-56645937 on Build GRCh38
rs866650910 Chr.12: 56645958-56645958 on Build GRCh38
rs530583169 Chr.12: 56645976-56645976 on Build GRCh38
rs779025464 Chr.12: 56645884-56645884 on Build GRCh38
rs757603602 Chr.12: 56645795-56645795 on Build GRCh38
rs769495213 Chr.12: 56645979-56645979 on Build GRCh38
rs11542646 Chr.12: 56645896-56645896 on Build GRCh38
rs748262122 Chr.12: 56645948-56645948 on Build GRCh38
rs748082322 Chr.12: 56645830-56645830 on Build GRCh38
rs755145018 Chr.12: 56645881-56645881 on Build GRCh38
rs559690374 Chr.12: 56645811-56645811 on Build GRCh38
rs577345165 Chr.12: 56645814-56645814 on Build GRCh38
rs767840117 Chr.12: 56645864-56645864 on Build GRCh38
rs776218742 Chr.12: 56645848-56645848 on Build GRCh38
rs776308441 Chr.12: 56645901-56645901 on Build GRCh38
rs969019085 Chr.12: 56645762-56645762 on Build GRCh38
rs780360695 Chr.12: 56645809-56645809 on Build GRCh38
rs749478722 Chr.12: 56645878-56645878 on Build GRCh38

View Full Product Details