Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 999563-999796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES4
Entrez Gene ID
57801
Gene Name
hes family bHLH transcription factor 4
Gene Aliases
bHLHb42
UniGene
Hs.154029
Gene Chromosome Location
Chr.1: 998962-1001052 on Build GRCh38
Transcripts
NM_001142467.1 NM_021170.3 XM_005244771.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352718 c.203T>C p.M68T Chr.1: 999771-999771 on Build GRCh38
4630453 c.162C>T p.L54L Chr.1: 999734-999734 on Build GRCh38
4848918 c.197C>G p.P66R Chr.1: 999777-999777 on Build GRCh38
4593509 c.208T>C p.S70P Chr.1: 999610-999610 on Build GRCh38
6679537 c.109-1G>A p.? Chr.1: 999788-999788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777546047 Chr.1: 999726-999726 on Build GRCh38
rs746586303 Chr.1: 999619-999619 on Build GRCh38
rs776060243 Chr.1: 999626-999626 on Build GRCh38
rs759783095 Chr.1: 999779-999779 on Build GRCh38
rs1040840388 Chr.1: 999622-999622 on Build GRCh38
rs190676096 Chr.1: 999756-999756 on Build GRCh38
rs777742231 Chr.1: 999680-999680 on Build GRCh38
rs370001003 Chr.1: 999657-999657 on Build GRCh38
rs751729184 Chr.1: 999587-999587 on Build GRCh38
rs972828069 Chr.1: 999774-999774 on Build GRCh38
rs756297814 Chr.1: 999599-999599 on Build GRCh38
rs771948328 Chr.1: 999611-999611 on Build GRCh38
rs531408504 Chr.1: 999701-999701 on Build GRCh38
rs776036679 Chr.1: 999689-999689 on Build GRCh38
rs768092304 Chr.1: 999712-999712 on Build GRCh38
rs747930284 Chr.1: 999672-999672 on Build GRCh38
rs369100513 Chr.1: 999652-999652 on Build GRCh38
rs375743015 Chr.1: 999640-999640 on Build GRCh38
rs968372114 Chr.1: 999704-999704 on Build GRCh38
rs763748416 Chr.1: 999689-999690 on Build GRCh38
rs764767141 Chr.1: 999638-999638 on Build GRCh38
rs775768823 Chr.1: 999635-999635 on Build GRCh38
rs750514369 Chr.1: 999713-999713 on Build GRCh38
rs144493256 Chr.1: 999709-999709 on Build GRCh38
rs201520566 Chr.1: 999610-999610 on Build GRCh38
rs780296768 Chr.1: 999671-999671 on Build GRCh38
rs781051403 Chr.1: 999748-999748 on Build GRCh38
rs757410292 Chr.1: 999588-999588 on Build GRCh38
rs148364701 Chr.1: 999621-999621 on Build GRCh38
rs781154824 Chr.1: 999732-999732 on Build GRCh38
rs751315962 Chr.1: 999730-999730 on Build GRCh38
rs140184340 Chr.1: 999720-999720 on Build GRCh38
rs773100525 Chr.1: 999612-999612 on Build GRCh38
rs754074223 Chr.1: 999725-999725 on Build GRCh38
rs762249677 Chr.1: 999754-999754 on Build GRCh38
rs770412889 Chr.1: 999620-999620 on Build GRCh38
rs758182930 Chr.1: 999679-999679 on Build GRCh38
rs764903126 Chr.1: 999629-999629 on Build GRCh38
rs950341015 Chr.1: 999605-999605 on Build GRCh38
rs775175678 Chr.1: 999616-999616 on Build GRCh38
rs920889968 Chr.1: 999590-999590 on Build GRCh38
rs370586563 Chr.1: 999707-999707 on Build GRCh38
rs753793334 Chr.1: 999770-999770 on Build GRCh38
rs1021682341 Chr.1: 999621-999621 on Build GRCh38
rs140166926 Chr.1: 999758-999758 on Build GRCh38
rs201579975 Chr.1: 999710-999710 on Build GRCh38
rs144665333 Chr.1: 999705-999705 on Build GRCh38
rs762668692 Chr.1: 999674-999674 on Build GRCh38
rs763346929 Chr.1: 999637-999637 on Build GRCh38
rs746961720 Chr.1: 999682-999682 on Build GRCh38
rs779375158 Chr.1: 999743-999743 on Build GRCh38
rs757207931 Chr.1: 999731-999731 on Build GRCh38
rs139739199 Chr.1: 999691-999691 on Build GRCh38
rs766490059 Chr.1: 999763-999763 on Build GRCh38
rs377588302 Chr.1: 999700-999700 on Build GRCh38
rs781624234 Chr.1: 999594-999594 on Build GRCh38
rs745436992 Chr.1: 999736-999736 on Build GRCh38
rs774882183 Chr.1: 999750-999750 on Build GRCh38
rs757783143 Chr.1: 999645-999645 on Build GRCh38
rs759115521 Chr.1: 999627-999627 on Build GRCh38
rs113602214 Chr.1: 999666-999666 on Build GRCh38
rs1049269510 Chr.1: 999661-999661 on Build GRCh38
rs746094252 Chr.1: 999595-999595 on Build GRCh38
rs1039053449 Chr.1: 999688-999688 on Build GRCh38
rs376997814 Chr.1: 999602-999602 on Build GRCh38
rs769335589 Chr.1: 999737-999737 on Build GRCh38
rs887846852 Chr.1: 999662-999662 on Build GRCh38
rs1000627427 Chr.1: 999719-999719 on Build GRCh38
rs111418382 Chr.1: 999757-999757 on Build GRCh38
rs959097937 Chr.1: 999721-999721 on Build GRCh38
rs760721734 Chr.1: 999762-999762 on Build GRCh38
rs568810432 Chr.1: 999685-999685 on Build GRCh38

View Full Product Details