Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17636411-17636672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184B
Entrez Gene ID
27146
Gene Name
family with sequence similarity 184 member B
Gene Aliases
-
UniGene
Hs.744951
Gene Chromosome Location
Chr.4: 17632073-17781512 on Build GRCh38
Transcripts
NM_015688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5975062 c.2772C>T p.I924I Chr.4: 17636540-17636540 on Build GRCh38
6314852 c.2746C>A p.R916S Chr.4: 17636566-17636566 on Build GRCh38
5771480 c.2667-3C>G p.? Chr.4: 17636648-17636648 on Build GRCh38
4780032 c.2690C>A p.P897Q Chr.4: 17636622-17636622 on Build GRCh38
6300881 c.2746C>T p.R916C Chr.4: 17636566-17636566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910177132 Chr.4: 17636609-17636609 on Build GRCh38
rs766723991 Chr.4: 17636494-17636494 on Build GRCh38
rs774679756 Chr.4: 17636627-17636627 on Build GRCh38
rs936058789 Chr.4: 17636554-17636554 on Build GRCh38
rs756490582 Chr.4: 17636479-17636479 on Build GRCh38
rs267600081 Chr.4: 17636599-17636599 on Build GRCh38
rs916459687 Chr.4: 17636515-17636515 on Build GRCh38
rs958930827 Chr.4: 17636614-17636614 on Build GRCh38
rs932158528 Chr.4: 17636618-17636618 on Build GRCh38
rs577846214 Chr.4: 17636452-17636452 on Build GRCh38
rs984690065 Chr.4: 17636607-17636607 on Build GRCh38
rs775369128 Chr.4: 17636590-17636590 on Build GRCh38
rs757988608 Chr.4: 17636510-17636510 on Build GRCh38
rs781050490 Chr.4: 17636561-17636561 on Build GRCh38
rs1041765368 Chr.4: 17636648-17636648 on Build GRCh38
rs774697465 Chr.4: 17636539-17636541 on Build GRCh38
rs746429072 Chr.4: 17636547-17636549 on Build GRCh38
rs746563665 Chr.4: 17636513-17636513 on Build GRCh38
rs979226818 Chr.4: 17636549-17636550 on Build GRCh38
rs777286657 Chr.4: 17636512-17636512 on Build GRCh38
rs766095565 Chr.4: 17636466-17636467 on Build GRCh38
rs1004571941 Chr.4: 17636498-17636498 on Build GRCh38
rs900660915 Chr.4: 17636480-17636480 on Build GRCh38
rs370226374 Chr.4: 17636577-17636577 on Build GRCh38
rs373439076 Chr.4: 17636588-17636588 on Build GRCh38
rs375831461 Chr.4: 17636568-17636568 on Build GRCh38
rs147570922 Chr.4: 17636519-17636520 on Build GRCh38
rs548562323 Chr.4: 17636470-17636470 on Build GRCh38
rs917220930 Chr.4: 17636633-17636633 on Build GRCh38
rs960685887 Chr.4: 17636509-17636509 on Build GRCh38
rs11944484 Chr.4: 17636555-17636555 on Build GRCh38
rs993216332 Chr.4: 17636578-17636578 on Build GRCh38
rs550537049 Chr.4: 17636622-17636622 on Build GRCh38
rs1032894120 Chr.4: 17636496-17636496 on Build GRCh38
rs149435417 Chr.4: 17636456-17636456 on Build GRCh38
rs1026249610 Chr.4: 17636595-17636595 on Build GRCh38
rs879121604 Chr.4: 17636547-17636547 on Build GRCh38
rs751630669 Chr.4: 17636488-17636488 on Build GRCh38
rs887549558 Chr.4: 17636654-17636654 on Build GRCh38
rs369519344 Chr.4: 17636508-17636508 on Build GRCh38
rs752349343 Chr.4: 17636499-17636499 on Build GRCh38
rs1041549754 Chr.4: 17636469-17636469 on Build GRCh38
rs530812460 Chr.4: 17636583-17636583 on Build GRCh38
rs749548028 Chr.4: 17636600-17636600 on Build GRCh38
rs1040601621 Chr.4: 17636613-17636613 on Build GRCh38
rs1014975650 Chr.4: 17636500-17636500 on Build GRCh38
rs570642756 Chr.4: 17636632-17636632 on Build GRCh38
rs950156225 Chr.4: 17636644-17636644 on Build GRCh38
rs1018602034 Chr.4: 17636566-17636566 on Build GRCh38
rs528497369 Chr.4: 17636461-17636461 on Build GRCh38
rs926027663 Chr.4: 17636553-17636553 on Build GRCh38
rs1040548367 Chr.4: 17636461-17636462 on Build GRCh38
rs778785791 Chr.4: 17636549-17636552 on Build GRCh38

View Full Product Details