Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 113944201-113944470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC2
Entrez Gene ID
3066
Gene Name
histone deacetylase 2
Gene Aliases
HD2, RPD3, YAF1
UniGene
Hs.3352
Gene Chromosome Location
Chr.6: 113936156-113971195 on Build GRCh38
Transcripts
NM_001527.3 NR_033441.1 NR_073443.1 XM_011535788.1 XM_017010799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559444 c.1112T>C p.L371S Chr.6: 113944390-113944390 on Build GRCh38
7286745 c.1395G>A p.L465L Chr.6: 113944389-113944389 on Build GRCh38
3156876 c.1125T>A p.P375P Chr.6: 113944377-113944377 on Build GRCh38
4460200 c.1438C>T p.P480S Chr.6: 113944346-113944346 on Build GRCh38
5559443 c.1394T>C p.L465S Chr.6: 113944390-113944390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750391829 Chr.6: 113944306-113944306 on Build GRCh38
rs761440609 Chr.6: 113944280-113944280 on Build GRCh38
rs375131198 Chr.6: 113944421-113944421 on Build GRCh38
rs989228615 Chr.6: 113944273-113944273 on Build GRCh38
rs192805602 Chr.6: 113944291-113944291 on Build GRCh38
rs964641741 Chr.6: 113944283-113944283 on Build GRCh38
rs377368556 Chr.6: 113944243-113944243 on Build GRCh38
rs13204445 Chr.6: 113944423-113944423 on Build GRCh38
rs556116783 Chr.6: 113944374-113944374 on Build GRCh38
rs763407740 Chr.6: 113944413-113944413 on Build GRCh38
rs375129341 Chr.6: 113944262-113944262 on Build GRCh38
rs758065478 Chr.6: 113944315-113944315 on Build GRCh38
rs368283587 Chr.6: 113944344-113944344 on Build GRCh38
rs957898651 Chr.6: 113944257-113944257 on Build GRCh38
rs777795421 Chr.6: 113944356-113944356 on Build GRCh38
rs116014206 Chr.6: 113944254-113944254 on Build GRCh38
rs184982628 Chr.6: 113944353-113944353 on Build GRCh38
rs780519196 Chr.6: 113944437-113944437 on Build GRCh38
rs76956103 Chr.6: 113944410-113944410 on Build GRCh38
rs200302916 Chr.6: 113944268-113944268 on Build GRCh38
rs1030351654 Chr.6: 113944237-113944237 on Build GRCh38
rs1019826364 Chr.6: 113944269-113944269 on Build GRCh38
rs774936510 Chr.6: 113944242-113944242 on Build GRCh38
rs765221258 Chr.6: 113944301-113944301 on Build GRCh38
rs772496066 Chr.6: 113944239-113944239 on Build GRCh38
rs766266910 Chr.6: 113944339-113944339 on Build GRCh38
rs772318119 Chr.6: 113944424-113944424 on Build GRCh38
rs966502661 Chr.6: 113944335-113944335 on Build GRCh38
rs746166272 Chr.6: 113944238-113944238 on Build GRCh38
rs753870308 Chr.6: 113944368-113944368 on Build GRCh38

View Full Product Details