Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52775443-52775682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT76
Entrez Gene ID
51350
Gene Name
keratin 76
Gene Aliases
HUMCYT2A, KRT2B, KRT2P
UniGene
Hs.654392
Gene Chromosome Location
Chr.12: 52768155-52777345 on Build GRCh38
Transcripts
NM_015848.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4911185 c.708C>A p.S236S Chr.12: 52775495-52775495 on Build GRCh38
1739936 c.658C>T p.Q220* Chr.12: 52775545-52775545 on Build GRCh38
4042920 c.610C>A p.L204M Chr.12: 52775593-52775593 on Build GRCh38
5533556 c.688C>T p.L230L Chr.12: 52775515-52775515 on Build GRCh38
6877475 c.707C>T p.S236F Chr.12: 52775496-52775496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780155117 Chr.12: 52775632-52775632 on Build GRCh38
rs61730601 Chr.12: 52775465-52775465 on Build GRCh38
rs758688800 Chr.12: 52775510-52775510 on Build GRCh38
rs559709444 Chr.12: 52775648-52775648 on Build GRCh38
rs750437723 Chr.12: 52775629-52775629 on Build GRCh38
rs374515260 Chr.12: 52775642-52775642 on Build GRCh38
rs140457873 Chr.12: 52775560-52775560 on Build GRCh38
rs772626759 Chr.12: 52775627-52775627 on Build GRCh38
rs780262137 Chr.12: 52775554-52775554 on Build GRCh38
rs765570274 Chr.12: 52775572-52775572 on Build GRCh38
rs147716256 Chr.12: 52775494-52775494 on Build GRCh38
rs766905459 Chr.12: 52775489-52775489 on Build GRCh38
rs912973113 Chr.12: 52775655-52775655 on Build GRCh38
rs761615359 Chr.12: 52775582-52775582 on Build GRCh38
rs149176912 Chr.12: 52775496-52775496 on Build GRCh38
rs781209244 Chr.12: 52775531-52775531 on Build GRCh38
rs757826922 Chr.12: 52775529-52775529 on Build GRCh38
rs761858227 Chr.12: 52775483-52775483 on Build GRCh38
rs768953368 Chr.12: 52775636-52775636 on Build GRCh38
rs370398922 Chr.12: 52775598-52775598 on Build GRCh38
rs772638016 Chr.12: 52775633-52775633 on Build GRCh38
rs768560961 Chr.12: 52775471-52775471 on Build GRCh38
rs143290453 Chr.12: 52775512-52775512 on Build GRCh38
rs766140677 Chr.12: 52775501-52775501 on Build GRCh38
rs751992504 Chr.12: 52775523-52775523 on Build GRCh38
rs750818000 Chr.12: 52775614-52775614 on Build GRCh38
rs753530688 Chr.12: 52775602-52775602 on Build GRCh38
rs759285235 Chr.12: 52775606-52775606 on Build GRCh38
rs774140731 Chr.12: 52775478-52775478 on Build GRCh38
rs781674257 Chr.12: 52775628-52775628 on Build GRCh38
rs768312111 Chr.12: 52775561-52775561 on Build GRCh38
rs374789954 Chr.12: 52775608-52775608 on Build GRCh38
rs1013327714 Chr.12: 52775645-52775645 on Build GRCh38
rs762320246 Chr.12: 52775552-52775552 on Build GRCh38
rs769948069 Chr.12: 52775545-52775545 on Build GRCh38
rs530359926 Chr.12: 52775654-52775654 on Build GRCh38
rs945852723 Chr.12: 52775658-52775658 on Build GRCh38
rs145628518 Chr.12: 52775599-52775599 on Build GRCh38
rs754643966 Chr.12: 52775619-52775622 on Build GRCh38
rs543446586 Chr.12: 52775466-52775466 on Build GRCh38
rs745970657 Chr.12: 52775537-52775537 on Build GRCh38
rs370873599 Chr.12: 52775630-52775630 on Build GRCh38
rs774241532 Chr.12: 52775566-52775566 on Build GRCh38
rs962189348 Chr.12: 52775577-52775577 on Build GRCh38
rs753041037 Chr.12: 52775509-52775509 on Build GRCh38
rs148316600 Chr.12: 52775525-52775525 on Build GRCh38
rs778520064 Chr.12: 52775637-52775637 on Build GRCh38
rs376112650 Chr.12: 52775623-52775623 on Build GRCh38
rs772065201 Chr.12: 52775595-52775595 on Build GRCh38

View Full Product Details