Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112149682-112149945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
UniGene
Hs.5148
Gene Chromosome Location
Chr.12: 112125545-112153604 on Build GRCh38
Transcripts
NM_001143906.1 NM_006700.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1706291 c.1234C>T p.Q412* Chr.12: 112149826-112149826 on Build GRCh38
4575026 c.1196A>G p.H399R Chr.12: 112149788-112149788 on Build GRCh38
6807296 c.1171C>A p.Q391K Chr.12: 112149763-112149763 on Build GRCh38
3954187 c.1234C>G p.Q412E Chr.12: 112149826-112149826 on Build GRCh38
3456159 c.1238A>C p.E413A Chr.12: 112149830-112149830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775760794 Chr.12: 112149808-112149808 on Build GRCh38
rs763124143 Chr.12: 112149855-112149855 on Build GRCh38
rs758289846 Chr.12: 112149736-112149736 on Build GRCh38
rs750184907 Chr.12: 112149735-112149735 on Build GRCh38
rs139453904 Chr.12: 112149922-112149922 on Build GRCh38
rs746185807 Chr.12: 112149796-112149796 on Build GRCh38
rs755477949 Chr.12: 112149909-112149929 on Build GRCh38
rs772401610 Chr.12: 112149703-112149703 on Build GRCh38
rs958553082 Chr.12: 112149871-112149871 on Build GRCh38
rs761993967 Chr.12: 112149848-112149848 on Build GRCh38
rs188071535 Chr.12: 112149867-112149867 on Build GRCh38
rs202109450 Chr.12: 112149829-112149829 on Build GRCh38
rs755648306 Chr.12: 112149889-112149889 on Build GRCh38
rs61941323 Chr.12: 112149894-112149894 on Build GRCh38
rs776087648 Chr.12: 112149705-112149705 on Build GRCh38
rs578177454 Chr.12: 112149834-112149834 on Build GRCh38
rs755697440 Chr.12: 112149781-112149781 on Build GRCh38
rs903073426 Chr.12: 112149837-112149837 on Build GRCh38
rs2293433 Chr.12: 112149921-112149921 on Build GRCh38
rs111354957 Chr.12: 112149713-112149713 on Build GRCh38
rs943859821 Chr.12: 112149903-112149903 on Build GRCh38
rs756099163 Chr.12: 112149852-112149852 on Build GRCh38
rs777613549 Chr.12: 112149856-112149856 on Build GRCh38
rs374232152 Chr.12: 112149849-112149849 on Build GRCh38
rs776630700 Chr.12: 112149716-112149716 on Build GRCh38
rs752315326 Chr.12: 112149711-112149711 on Build GRCh38
rs1035942152 Chr.12: 112149764-112149764 on Build GRCh38
rs562930540 Chr.12: 112149820-112149820 on Build GRCh38
rs765560758 Chr.12: 112149720-112149720 on Build GRCh38
rs766186145 Chr.12: 112149739-112149739 on Build GRCh38
rs777420303 Chr.12: 112149788-112149788 on Build GRCh38
rs747724400 Chr.12: 112149772-112149772 on Build GRCh38
rs751482095 Chr.12: 112149745-112149745 on Build GRCh38
rs754921888 Chr.12: 112149748-112149748 on Build GRCh38
rs759514302 Chr.12: 112149857-112149857 on Build GRCh38
rs369585563 Chr.12: 112149914-112149914 on Build GRCh38
rs985594093 Chr.12: 112149879-112149879 on Build GRCh38
rs760632836 Chr.12: 112149706-112149706 on Build GRCh38
rs749061357 Chr.12: 112149880-112149880 on Build GRCh38
rs780742090 Chr.12: 112149767-112149767 on Build GRCh38
rs753461637 Chr.12: 112149902-112149902 on Build GRCh38
rs764205528 Chr.12: 112149714-112149714 on Build GRCh38
rs963137944 Chr.12: 112149910-112149910 on Build GRCh38
rs956278519 Chr.12: 112149811-112149811 on Build GRCh38
rs772520742 Chr.12: 112149805-112149805 on Build GRCh38
rs560665923 Chr.12: 112149901-112149901 on Build GRCh38
rs756911235 Chr.12: 112149905-112149905 on Build GRCh38
rs762120868 Chr.12: 112149717-112149717 on Build GRCh38
rs376713214 Chr.12: 112149884-112149884 on Build GRCh38

View Full Product Details