Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42145953-42146180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G4F
Entrez Gene ID
255189
Gene Name
phospholipase A2 group IVF
Gene Aliases
PLA2G4FZ
UniGene
Hs.231873
Gene Chromosome Location
Chr.15: 42141134-42156641 on Build GRCh38
Transcripts
NM_213600.3 NR_033151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433858 c.1531G>T p.A511S Chr.15: 42146130-42146130 on Build GRCh38
5383301 c.1503C>T p.V501V Chr.15: 42146158-42146158 on Build GRCh38
7149508 c.1515G>C p.L505F Chr.15: 42146146-42146146 on Build GRCh38
6330742 c.1496T>A p.V499D Chr.15: 42146165-42146165 on Build GRCh38
6747759 c.1504C>T p.R502C Chr.15: 42146157-42146157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199839163 Chr.15: 42146144-42146144 on Build GRCh38
rs770080877 Chr.15: 42146146-42146146 on Build GRCh38
rs780556501 Chr.15: 42146114-42146114 on Build GRCh38
rs375514932 Chr.15: 42146099-42146099 on Build GRCh38
rs922156107 Chr.15: 42146056-42146056 on Build GRCh38
rs371560229 Chr.15: 42146127-42146127 on Build GRCh38
rs765605608 Chr.15: 42146136-42146136 on Build GRCh38
rs902827515 Chr.15: 42146048-42146048 on Build GRCh38
rs924984793 Chr.15: 42145985-42145985 on Build GRCh38
rs377552532 Chr.15: 42146150-42146150 on Build GRCh38
rs756985500 Chr.15: 42146093-42146093 on Build GRCh38
rs373729167 Chr.15: 42146139-42146139 on Build GRCh38
rs555310848 Chr.15: 42145996-42145996 on Build GRCh38
rs76858307 Chr.15: 42146109-42146109 on Build GRCh38
rs773236741 Chr.15: 42146152-42146152 on Build GRCh38
rs1049501273 Chr.15: 42146069-42146069 on Build GRCh38
rs747577617 Chr.15: 42146118-42146118 on Build GRCh38
rs746054219 Chr.15: 42146001-42146001 on Build GRCh38
rs144626679 Chr.15: 42146156-42146156 on Build GRCh38
rs753255715 Chr.15: 42146090-42146090 on Build GRCh38
rs932266487 Chr.15: 42146065-42146065 on Build GRCh38
rs887789756 Chr.15: 42146084-42146084 on Build GRCh38
rs114152163 Chr.15: 42146087-42146087 on Build GRCh38
rs749438331 Chr.15: 42146005-42146005 on Build GRCh38
rs1036397820 Chr.15: 42146097-42146097 on Build GRCh38
rs746275320 Chr.15: 42146108-42146108 on Build GRCh38
rs542774703 Chr.15: 42146028-42146028 on Build GRCh38
rs752623713 Chr.15: 42146078-42146078 on Build GRCh38
rs573776764 Chr.15: 42146009-42146009 on Build GRCh38
rs772537730 Chr.15: 42146082-42146095 on Build GRCh38
rs777961094 Chr.15: 42145974-42145974 on Build GRCh38
rs775997219 Chr.15: 42146087-42146095 on Build GRCh38
rs1018638819 Chr.15: 42146021-42146021 on Build GRCh38
rs956624089 Chr.15: 42145990-42145990 on Build GRCh38
rs151028116 Chr.15: 42146157-42146157 on Build GRCh38
rs769149240 Chr.15: 42146129-42146129 on Build GRCh38
rs779526342 Chr.15: 42146022-42146022 on Build GRCh38
rs971686573 Chr.15: 42146122-42146122 on Build GRCh38

View Full Product Details