Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102891194-102891467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF3
Entrez Gene ID
7187
Gene Name
TNF receptor associated factor 3
Gene Aliases
CAP-1, CAP1, CD40bp, CRAF1, IIAE5, LAP1
UniGene
Hs.510528
Gene Chromosome Location
Chr.14: 102777479-102911500 on Build GRCh38
Transcripts
NM_001199427.1 NM_003300.3 NM_145725.2 NM_145726.2 XM_011537116.2 XM_011537117.2 XM_011537118.2 XM_017021617.1 XM_017021618.1 XM_017021619.1 XM_017021620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944030 c.815_815delA p.K272fs*11 Chr.14: 102891413-102891413 on Build GRCh38
5504080 c.730A>G p.T244A Chr.14: 102891328-102891328 on Build GRCh38
5462356 c.819G>T p.K273N Chr.14: 102891417-102891417 on Build GRCh38
5053802 c.778G>A p.V260I Chr.14: 102891376-102891376 on Build GRCh38
6807222 c.810C>T p.L270L Chr.14: 102891408-102891408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755678092 Chr.14: 102891327-102891327 on Build GRCh38
rs749849268 Chr.14: 102891412-102891412 on Build GRCh38
rs2273393 Chr.14: 102891266-102891266 on Build GRCh38
rs200177742 Chr.14: 102891421-102891421 on Build GRCh38
rs376056808 Chr.14: 102891429-102891429 on Build GRCh38
rs753624346 Chr.14: 102891333-102891333 on Build GRCh38
rs767042790 Chr.14: 102891411-102891411 on Build GRCh38
rs376963322 Chr.14: 102891318-102891318 on Build GRCh38
rs944037805 Chr.14: 102891254-102891254 on Build GRCh38
rs149327017 Chr.14: 102891328-102891328 on Build GRCh38
rs895594487 Chr.14: 102891399-102891399 on Build GRCh38
rs771149972 Chr.14: 102891367-102891367 on Build GRCh38
rs938332234 Chr.14: 102891255-102891255 on Build GRCh38
rs529843465 Chr.14: 102891224-102891224 on Build GRCh38
rs543415634 Chr.14: 102891269-102891269 on Build GRCh38
rs746133902 Chr.14: 102891375-102891375 on Build GRCh38
rs757652604 Chr.14: 102891366-102891366 on Build GRCh38
rs778468210 Chr.14: 102891308-102891308 on Build GRCh38
rs751256024 Chr.14: 102891243-102891243 on Build GRCh38
rs763539329 Chr.14: 102891288-102891288 on Build GRCh38
rs769860474 Chr.14: 102891376-102891376 on Build GRCh38
rs759232349 Chr.14: 102891244-102891244 on Build GRCh38
rs750091557 Chr.14: 102891321-102891321 on Build GRCh38
rs891528259 Chr.14: 102891265-102891265 on Build GRCh38
rs755666441 Chr.14: 102891294-102891294 on Build GRCh38
rs775415215 Chr.14: 102891288-102891289 on Build GRCh38
rs754876308 Chr.14: 102891430-102891430 on Build GRCh38
rs767004037 Chr.14: 102891320-102891320 on Build GRCh38
rs909048499 Chr.14: 102891233-102891233 on Build GRCh38
rs144578195 Chr.14: 102891372-102891372 on Build GRCh38
rs767276346 Chr.14: 102891245-102891245 on Build GRCh38
rs752034706 Chr.14: 102891295-102891295 on Build GRCh38
rs779058785 Chr.14: 102891363-102891363 on Build GRCh38
rs763271165 Chr.14: 102891400-102891400 on Build GRCh38
rs765987424 Chr.14: 102891424-102891424 on Build GRCh38
rs754770677 Chr.14: 102891355-102891355 on Build GRCh38
rs560306318 Chr.14: 102891309-102891309 on Build GRCh38
rs775762527 Chr.14: 102891380-102891380 on Build GRCh38
rs752687128 Chr.14: 102891440-102891440 on Build GRCh38
rs369640883 Chr.14: 102891404-102891404 on Build GRCh38
rs913472625 Chr.14: 102891286-102891286 on Build GRCh38
rs201211273 Chr.14: 102891405-102891405 on Build GRCh38
rs765065668 Chr.14: 102891433-102891433 on Build GRCh38
rs1052781899 Chr.14: 102891262-102891262 on Build GRCh38
rs912538863 Chr.14: 102891246-102891246 on Build GRCh38

View Full Product Details