Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70324780-70325053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIA1
Entrez Gene ID
8500
Gene Name
PTPRF interacting protein alpha 1
Gene Aliases
LIP.1, LIP1, LIPRIN
UniGene
Hs.530749
Gene Chromosome Location
Chr.11: 70270687-70384501 on Build GRCh38
Transcripts
NM_003626.3 NM_177423.2 NR_045286.1 XM_006718716.3 XM_011545306.2 XM_011545307.2 XM_011545308.2 XM_011545309.2 XM_011545310.2 XM_011545311.2 XM_011545312.2 XM_011545313.2 XM_011545314.2 XM_011545315.2 XM_011545316.2 XM_011545317.2 XM_011545319.1 XM_017018448.1 XM_017018449.1 XM_017018450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4716861 c.500T>G p.L167* Chr.11: 70324980-70324980 on Build GRCh38
78330 c.410G>A p.R137Q Chr.11: 70324890-70324890 on Build GRCh38
1475849 c.474G>T p.V158V Chr.11: 70324954-70324954 on Build GRCh38
1475850 c.483G>C p.L161L Chr.11: 70324963-70324963 on Build GRCh38
931451 c.458G>A p.G153D Chr.11: 70324938-70324938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764425609 Chr.11: 70324889-70324889 on Build GRCh38
rs745973350 Chr.11: 70324965-70324965 on Build GRCh38
rs771977390 Chr.11: 70324838-70324838 on Build GRCh38
rs761644120 Chr.11: 70324834-70324834 on Build GRCh38
rs769856790 Chr.11: 70325020-70325020 on Build GRCh38
rs775743432 Chr.11: 70325031-70325031 on Build GRCh38
rs774499572 Chr.11: 70324828-70324828 on Build GRCh38
rs148677249 Chr.11: 70324939-70324939 on Build GRCh38
rs111283320 Chr.11: 70324931-70324931 on Build GRCh38
rs879794411 Chr.11: 70324948-70324948 on Build GRCh38
rs755632385 Chr.11: 70324924-70324924 on Build GRCh38
rs759612054 Chr.11: 70324857-70324857 on Build GRCh38
rs768539692 Chr.11: 70324817-70324817 on Build GRCh38
rs371918990 Chr.11: 70324901-70324901 on Build GRCh38
rs765491741 Chr.11: 70324861-70324861 on Build GRCh38
rs969145598 Chr.11: 70324934-70324934 on Build GRCh38
rs968740236 Chr.11: 70324835-70324835 on Build GRCh38
rs763371043 Chr.11: 70324885-70324885 on Build GRCh38
rs779894257 Chr.11: 70324929-70324929 on Build GRCh38
rs771923824 Chr.11: 70324956-70324956 on Build GRCh38
rs773179632 Chr.11: 70324963-70324963 on Build GRCh38
rs541397539 Chr.11: 70324930-70324930 on Build GRCh38
rs752881054 Chr.11: 70324870-70324870 on Build GRCh38
rs776603836 Chr.11: 70324844-70324844 on Build GRCh38
rs369798711 Chr.11: 70324888-70324888 on Build GRCh38
rs959183964 Chr.11: 70324937-70324937 on Build GRCh38
rs781696901 Chr.11: 70324913-70324913 on Build GRCh38
rs749928660 Chr.11: 70324923-70324923 on Build GRCh38
rs144282210 Chr.11: 70324906-70324906 on Build GRCh38
rs763790956 Chr.11: 70324834-70324837 on Build GRCh38
rs757711997 Chr.11: 70324907-70324907 on Build GRCh38
rs756011012 Chr.11: 70324940-70324940 on Build GRCh38

View Full Product Details