Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46588018-46588279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGF
Entrez Gene ID
5281
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Gene Aliases
-
UniGene
Hs.468415
Gene Chromosome Location
Chr.2: 46581274-46617112 on Build GRCh38
Transcripts
NM_002643.3 NM_173074.2 XM_011532908.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7121614 c.578G>C p.R193T Chr.2: 46588148-46588148 on Build GRCh38
4446826 c.547-3C>T p.? Chr.2: 46588182-46588182 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375801333 Chr.2: 46588209-46588209 on Build GRCh38
rs760063811 Chr.2: 46588146-46588146 on Build GRCh38
rs748298713 Chr.2: 46588222-46588222 on Build GRCh38
rs761148992 Chr.2: 46588139-46588139 on Build GRCh38
rs773273840 Chr.2: 46588138-46588138 on Build GRCh38
rs748338430 Chr.2: 46588132-46588132 on Build GRCh38
rs76065703 Chr.2: 46588077-46588077 on Build GRCh38
rs11125083 Chr.2: 46588085-46588085 on Build GRCh38
rs192397780 Chr.2: 46588150-46588150 on Build GRCh38
rs756008975 Chr.2: 46588174-46588174 on Build GRCh38
rs111331682 Chr.2: 46588178-46588178 on Build GRCh38
rs762025747 Chr.2: 46588058-46588058 on Build GRCh38
rs746671787 Chr.2: 46588081-46588081 on Build GRCh38
rs754858821 Chr.2: 46588196-46588196 on Build GRCh38
rs755844484 Chr.2: 46588107-46588107 on Build GRCh38
rs112738547 Chr.2: 46588045-46588045 on Build GRCh38
rs767784266 Chr.2: 46588060-46588060 on Build GRCh38
rs375421420 Chr.2: 46588137-46588137 on Build GRCh38
rs745719159 Chr.2: 46588106-46588106 on Build GRCh38
rs867209709 Chr.2: 46588075-46588075 on Build GRCh38
rs187929990 Chr.2: 46588064-46588064 on Build GRCh38
rs751270952 Chr.2: 46588089-46588089 on Build GRCh38
rs767252203 Chr.2: 46588161-46588161 on Build GRCh38
rs888843578 Chr.2: 46588195-46588195 on Build GRCh38
rs758398761 Chr.2: 46588062-46588063 on Build GRCh38
rs774353659 Chr.2: 46588125-46588125 on Build GRCh38
rs116004162 Chr.2: 46588156-46588156 on Build GRCh38
rs771436468 Chr.2: 46588225-46588225 on Build GRCh38
rs778114295 Chr.2: 46588224-46588224 on Build GRCh38
rs761116580 Chr.2: 46588066-46588066 on Build GRCh38
rs368263327 Chr.2: 46588061-46588061 on Build GRCh38
rs966684813 Chr.2: 46588129-46588129 on Build GRCh38
rs780882798 Chr.2: 46588099-46588099 on Build GRCh38
rs753784686 Chr.2: 46588187-46588187 on Build GRCh38
rs768723801 Chr.2: 46588122-46588122 on Build GRCh38
rs766836733 Chr.2: 46588142-46588142 on Build GRCh38
rs577117560 Chr.2: 46588084-46588084 on Build GRCh38
rs777281825 Chr.2: 46588144-46588144 on Build GRCh38
rs372568734 Chr.2: 46588160-46588160 on Build GRCh38
rs750034917 Chr.2: 46588171-46588171 on Build GRCh38
rs374861008 Chr.2: 46588080-46588080 on Build GRCh38
rs780697216 Chr.2: 46588119-46588119 on Build GRCh38
rs779754666 Chr.2: 46588181-46588181 on Build GRCh38
rs772022000 Chr.2: 46588223-46588223 on Build GRCh38
rs780028708 Chr.2: 46588113-46588113 on Build GRCh38
rs867638745 Chr.2: 46588078-46588078 on Build GRCh38

View Full Product Details