Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49220116-49220355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI3
Entrez Gene ID
51225
Gene Name
ABI family member 3
Gene Aliases
NESH, SSH3BP3
UniGene
Hs.130719
Gene Chromosome Location
Chr.17: 49210227-49223225 on Build GRCh38
Transcripts
NM_001135186.1 NM_016428.2 XM_005257429.2 XM_011524873.1 XM_017024721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147361 c.645C>T p.G215G Chr.17: 49220169-49220169 on Build GRCh38
1710447 c.728C>T p.P243L Chr.17: 49220252-49220252 on Build GRCh38
3518904 c.670C>T p.P224S Chr.17: 49220194-49220194 on Build GRCh38
1710446 c.682G>A p.G228R Chr.17: 49220206-49220206 on Build GRCh38
6590978 c.716G>T p.S239I Chr.17: 49220240-49220240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376591517 Chr.17: 49220190-49220190 on Build GRCh38
rs748908202 Chr.17: 49220226-49220226 on Build GRCh38
rs760800624 Chr.17: 49220241-49220241 on Build GRCh38
rs972914652 Chr.17: 49220180-49220180 on Build GRCh38
rs760290407 Chr.17: 49220149-49220149 on Build GRCh38
rs530493979 Chr.17: 49220320-49220320 on Build GRCh38
rs768810295 Chr.17: 49220290-49220290 on Build GRCh38
rs369387027 Chr.17: 49220151-49220151 on Build GRCh38
rs147987389 Chr.17: 49220169-49220169 on Build GRCh38
rs201222048 Chr.17: 49220159-49220159 on Build GRCh38
rs770587088 Chr.17: 49220272-49220272 on Build GRCh38
rs752155600 Chr.17: 49220315-49220315 on Build GRCh38
rs764364159 Chr.17: 49220198-49220198 on Build GRCh38
rs954284376 Chr.17: 49220139-49220139 on Build GRCh38
rs754487954 Chr.17: 49220204-49220204 on Build GRCh38
rs548270471 Chr.17: 49220259-49220259 on Build GRCh38
rs757687924 Chr.17: 49220199-49220199 on Build GRCh38
rs766971815 Chr.17: 49220314-49220314 on Build GRCh38
rs774120052 Chr.17: 49220227-49220227 on Build GRCh38
rs765454069 Chr.17: 49220298-49220298 on Build GRCh38
rs746478318 Chr.17: 49220144-49220144 on Build GRCh38
rs779628203 Chr.17: 49220150-49220150 on Build GRCh38
rs747823680 Chr.17: 49220153-49220153 on Build GRCh38
rs749017262 Chr.17: 49220221-49220221 on Build GRCh38
rs777262880 Chr.17: 49220262-49220262 on Build GRCh38
rs76617033 Chr.17: 49220175-49220175 on Build GRCh38
rs375202610 Chr.17: 49220274-49220274 on Build GRCh38
rs764197907 Chr.17: 49220242-49220242 on Build GRCh38
rs1046275945 Chr.17: 49220270-49220270 on Build GRCh38
rs756329744 Chr.17: 49220191-49220191 on Build GRCh38
rs762107097 Chr.17: 49220296-49220296 on Build GRCh38
rs139539456 Chr.17: 49220236-49220236 on Build GRCh38
rs760712996 Chr.17: 49220285-49220285 on Build GRCh38
rs750812549 Chr.17: 49220302-49220302 on Build GRCh38
rs750836534 Chr.17: 49220253-49220253 on Build GRCh38
rs150532080 Chr.17: 49220200-49220200 on Build GRCh38
rs777540796 Chr.17: 49220220-49220220 on Build GRCh38
rs979191054 Chr.17: 49220291-49220291 on Build GRCh38
rs775453030 Chr.17: 49220276-49220276 on Build GRCh38
rs368043560 Chr.17: 49220243-49220243 on Build GRCh38
rs747735844 Chr.17: 49220208-49220208 on Build GRCh38
rs528296462 Chr.17: 49220254-49220254 on Build GRCh38
rs772068630 Chr.17: 49220231-49220231 on Build GRCh38
rs778121089 Chr.17: 49220136-49220136 on Build GRCh38
rs369790283 Chr.17: 49220206-49220206 on Build GRCh38
rs141746983 Chr.17: 49220295-49220295 on Build GRCh38
rs763471257 Chr.17: 49220310-49220310 on Build GRCh38
rs200867869 Chr.17: 49220185-49220185 on Build GRCh38
rs771980419 Chr.17: 49220275-49220275 on Build GRCh38
rs759494930 Chr.17: 49220230-49220230 on Build GRCh38
rs749732235 Chr.17: 49220137-49220137 on Build GRCh38
rs999798939 Chr.17: 49220287-49220287 on Build GRCh38
rs376071536 Chr.17: 49220143-49220143 on Build GRCh38
rs763462624 Chr.17: 49220175-49220176 on Build GRCh38
rs371508417 Chr.17: 49220255-49220255 on Build GRCh38
rs752243727 Chr.17: 49220257-49220257 on Build GRCh38
rs769567980 Chr.17: 49220210-49220210 on Build GRCh38
rs749003957 Chr.17: 49220271-49220271 on Build GRCh38
rs755541670 Chr.17: 49220325-49220325 on Build GRCh38
rs757697687 Chr.17: 49220138-49220138 on Build GRCh38
rs368292072 Chr.17: 49220333-49220333 on Build GRCh38
rs765656420 Chr.17: 49220251-49220251 on Build GRCh38
rs559234442 Chr.17: 49220176-49220176 on Build GRCh38
rs1015906035 Chr.17: 49220189-49220189 on Build GRCh38
rs993075248 Chr.17: 49220249-49220249 on Build GRCh38

View Full Product Details