Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64826372-64826631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPG
Entrez Gene ID
55561
Gene Name
CDC42 binding protein kinase gamma
Gene Aliases
DMPK2, HSMDPKIN, KAPPA-200, MRCKG, MRCKgamma
UniGene
Hs.293590
Gene Chromosome Location
Chr.11: 64823809-64844685 on Build GRCh38
Transcripts
NM_017525.2 XM_011545155.2 XM_011545156.2 XM_011545157.2 XM_011545158.2 XM_011545159.2 XM_011545160.2 XM_017017996.1 XM_017017999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451587 c.4550C>T p.S1517F Chr.11: 64826519-64826519 on Build GRCh38
6860048 c.4539G>A p.L1513L Chr.11: 64826530-64826530 on Build GRCh38
3451586 c.4563C>T p.P1521P Chr.11: 64826506-64826506 on Build GRCh38
2164515 c.4536C>T p.S1512S Chr.11: 64826533-64826533 on Build GRCh38
5480917 c.4531A>G p.T1511A Chr.11: 64826538-64826538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756825767 Chr.11: 64826453-64826453 on Build GRCh38
rs962654418 Chr.11: 64826459-64826459 on Build GRCh38
rs908639259 Chr.11: 64826542-64826542 on Build GRCh38
rs370668615 Chr.11: 64826464-64826464 on Build GRCh38
rs375458071 Chr.11: 64826549-64826549 on Build GRCh38
rs373268624 Chr.11: 64826452-64826452 on Build GRCh38
rs200346114 Chr.11: 64826451-64826451 on Build GRCh38
rs975191908 Chr.11: 64826608-64826608 on Build GRCh38
rs983942515 Chr.11: 64826532-64826532 on Build GRCh38
rs760002344 Chr.11: 64826533-64826533 on Build GRCh38
rs199564155 Chr.11: 64826567-64826567 on Build GRCh38
rs373481886 Chr.11: 64826423-64826423 on Build GRCh38
rs373118105 Chr.11: 64826492-64826492 on Build GRCh38
rs377424840 Chr.11: 64826503-64826503 on Build GRCh38
rs776945735 Chr.11: 64826527-64826527 on Build GRCh38
rs1056163237 Chr.11: 64826440-64826440 on Build GRCh38
rs71526500 Chr.11: 64826457-64826457 on Build GRCh38
rs771470342 Chr.11: 64826524-64826524 on Build GRCh38
rs761238022 Chr.11: 64826430-64826430 on Build GRCh38
rs760628500 Chr.11: 64826523-64826523 on Build GRCh38
rs893980121 Chr.11: 64826417-64826417 on Build GRCh38
rs761434226 Chr.11: 64826559-64826559 on Build GRCh38
rs191214442 Chr.11: 64826570-64826570 on Build GRCh38
rs868258731 Chr.11: 64826482-64826482 on Build GRCh38
rs759753129 Chr.11: 64826448-64826448 on Build GRCh38
rs753709120 Chr.11: 64826575-64826575 on Build GRCh38
rs558837870 Chr.11: 64826506-64826506 on Build GRCh38
rs778825991 Chr.11: 64826576-64826576 on Build GRCh38
rs995109398 Chr.11: 64826470-64826470 on Build GRCh38
rs574134052 Chr.11: 64826588-64826588 on Build GRCh38
rs747125782 Chr.11: 64826596-64826596 on Build GRCh38
rs555841078 Chr.11: 64826399-64826399 on Build GRCh38
rs756420531 Chr.11: 64826474-64826475 on Build GRCh38
rs369700316 Chr.11: 64826450-64826450 on Build GRCh38
rs750411179 Chr.11: 64826461-64826461 on Build GRCh38
rs768281378 Chr.11: 64826497-64826497 on Build GRCh38
rs375949928 Chr.11: 64826437-64826437 on Build GRCh38
rs767109978 Chr.11: 64826565-64826565 on Build GRCh38
rs766744575 Chr.11: 64826435-64826435 on Build GRCh38
rs1042663532 Chr.11: 64826393-64826393 on Build GRCh38
rs959693660 Chr.11: 64826485-64826485 on Build GRCh38
rs370418079 Chr.11: 64826573-64826573 on Build GRCh38
rs765513496 Chr.11: 64826548-64826548 on Build GRCh38
rs922636983 Chr.11: 64826573-64826575 on Build GRCh38
rs747899315 Chr.11: 64826579-64826579 on Build GRCh38
rs573460035 Chr.11: 64826426-64826426 on Build GRCh38
rs370423505 Chr.11: 64826505-64826505 on Build GRCh38
rs780139587 Chr.11: 64826476-64826477 on Build GRCh38

View Full Product Details