Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42562376-42562637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COASY
Entrez Gene ID
80347
Gene Name
Coenzyme A synthase
Gene Aliases
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2
UniGene
Hs.296422
Gene Chromosome Location
Chr.17: 42562074-42566281 on Build GRCh38
Transcripts
NM_001042529.2 NM_001042532.3 NM_025233.6 XM_006722116.3 XM_011525300.1 XM_017025167.1 XM_017025168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893915 c.70C>T p.P24S Chr.17: 42562481-42562481 on Build GRCh38
5453094 c.6C>T p.A2A Chr.17: 42562628-42562628 on Build GRCh38
5453093 c.93C>T p.A31A Chr.17: 42562628-42562628 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370095840 Chr.17: 42562450-42562450 on Build GRCh38
rs949597584 Chr.17: 42562597-42562597 on Build GRCh38
rs1046803305 Chr.17: 42562582-42562582 on Build GRCh38
rs528235676 Chr.17: 42562447-42562447 on Build GRCh38
rs779309874 Chr.17: 42562606-42562606 on Build GRCh38
rs919235639 Chr.17: 42562424-42562424 on Build GRCh38
rs763334155 Chr.17: 42562403-42562403 on Build GRCh38
rs769149059 Chr.17: 42562465-42562465 on Build GRCh38
rs892158923 Chr.17: 42562580-42562580 on Build GRCh38
rs755354716 Chr.17: 42562573-42562573 on Build GRCh38
rs759055031 Chr.17: 42562570-42562570 on Build GRCh38
rs773425303 Chr.17: 42562522-42562522 on Build GRCh38
rs201479820 Chr.17: 42562535-42562535 on Build GRCh38
rs774636354 Chr.17: 42562467-42562467 on Build GRCh38
rs370287461 Chr.17: 42562583-42562583 on Build GRCh38
rs2301132 Chr.17: 42562507-42562507 on Build GRCh38
rs766034518 Chr.17: 42562476-42562476 on Build GRCh38
rs763993868 Chr.17: 42562594-42562594 on Build GRCh38
rs762780076 Chr.17: 42562591-42562591 on Build GRCh38
rs986711026 Chr.17: 42562479-42562479 on Build GRCh38
rs766909925 Chr.17: 42562502-42562502 on Build GRCh38
rs772880365 Chr.17: 42562474-42562474 on Build GRCh38
rs919671111 Chr.17: 42562561-42562561 on Build GRCh38
rs760282616 Chr.17: 42562475-42562475 on Build GRCh38
rs563959731 Chr.17: 42562400-42562400 on Build GRCh38
rs561601439 Chr.17: 42562483-42562483 on Build GRCh38
rs748492085 Chr.17: 42562511-42562511 on Build GRCh38
rs754940764 Chr.17: 42562442-42562442 on Build GRCh38
rs766448525 Chr.17: 42562440-42562440 on Build GRCh38
rs749835853 Chr.17: 42562463-42562463 on Build GRCh38
rs750696357 Chr.17: 42562497-42562497 on Build GRCh38
rs759611664 Chr.17: 42562482-42562482 on Build GRCh38
rs779409128 Chr.17: 42562508-42562508 on Build GRCh38
rs758442645 Chr.17: 42562472-42562472 on Build GRCh38
rs760441921 Chr.17: 42562528-42562528 on Build GRCh38
rs767186759 Chr.17: 42562422-42562422 on Build GRCh38
rs748736278 Chr.17: 42562607-42562607 on Build GRCh38
rs7219522 Chr.17: 42562531-42562531 on Build GRCh38
rs762180459 Chr.17: 42562471-42562471 on Build GRCh38
rs779997818 Chr.17: 42562537-42562537 on Build GRCh38
rs752442631 Chr.17: 42562485-42562485 on Build GRCh38
rs753921053 Chr.17: 42562602-42562602 on Build GRCh38
rs772431211 Chr.17: 42562512-42562512 on Build GRCh38
rs200710685 Chr.17: 42562490-42562490 on Build GRCh38
rs191068578 Chr.17: 42562460-42562460 on Build GRCh38
rs761439725 Chr.17: 42562419-42562419 on Build GRCh38
rs745969255 Chr.17: 42562461-42562461 on Build GRCh38
rs774748328 Chr.17: 42562417-42562417 on Build GRCh38
rs753365072 Chr.17: 42562478-42562478 on Build GRCh38
rs764492939 Chr.17: 42562414-42562414 on Build GRCh38
rs753779026 Chr.17: 42562441-42562441 on Build GRCh38
rs749914439 Chr.17: 42562425-42562425 on Build GRCh38
rs781294581 Chr.17: 42562492-42562492 on Build GRCh38
rs771627280 Chr.17: 42562472-42562472 on Build GRCh38
rs755619184 Chr.17: 42562429-42562429 on Build GRCh38
rs751353009 Chr.17: 42562596-42562596 on Build GRCh38
rs554944700 Chr.17: 42562613-42562613 on Build GRCh38
rs769818125 Chr.17: 42562462-42562462 on Build GRCh38
rs373992537 Chr.17: 42562604-42562604 on Build GRCh38
rs1032408401 Chr.17: 42562397-42562397 on Build GRCh38
rs780177247 Chr.17: 42562504-42562504 on Build GRCh38
rs780191999 Chr.17: 42562601-42562601 on Build GRCh38

View Full Product Details