Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 51698221-51698490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF2T
Entrez Gene ID
23283
Gene Name
cleavage stimulation factor subunit 2, tau variant
Gene Aliases
CstF-64T
UniGene
Hs.591358
Gene Chromosome Location
Chr.10: 51695486-51699595 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Gene Symbol
PRKG1
Entrez Gene ID
5592
Gene Name
protein kinase, cGMP-dependent, type I
Gene Aliases
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha
UniGene
Hs.407535
Gene Chromosome Location
Chr.10: 50991151-52298350 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4443787 c.1307G>A p.R436K Chr.10: 51698243-51698243 on Build GRCh38
3375482 c.1069C>T p.P357S Chr.10: 51698481-51698481 on Build GRCh38
1470425 c.1276G>A p.E426K Chr.10: 51698274-51698274 on Build GRCh38
5518974 c.1075delC p.H359fs*22 Chr.10: 51698475-51698475 on Build GRCh38
146938 c.1263T>C p.R421R Chr.10: 51698287-51698287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764346235 Chr.10: 51698432-51698432 on Build GRCh38
rs771847513 Chr.10: 51698288-51698288 on Build GRCh38
rs3824686 Chr.10: 51698407-51698407 on Build GRCh38
rs937252250 Chr.10: 51698285-51698285 on Build GRCh38
rs762563728 Chr.10: 51698310-51698318 on Build GRCh38
rs750851950 Chr.10: 51698267-51698267 on Build GRCh38
rs146369924 Chr.10: 51698282-51698282 on Build GRCh38
rs3740228 Chr.10: 51698287-51698287 on Build GRCh38
rs758806447 Chr.10: 51698272-51698272 on Build GRCh38
rs768087095 Chr.10: 51698293-51698293 on Build GRCh38
rs1036729694 Chr.10: 51698323-51698323 on Build GRCh38
rs1003268596 Chr.10: 51698358-51698358 on Build GRCh38
rs773013114 Chr.10: 51698434-51698434 on Build GRCh38
rs749495404 Chr.10: 51698386-51698386 on Build GRCh38
rs963653879 Chr.10: 51698351-51698351 on Build GRCh38
rs765545916 Chr.10: 51698263-51698263 on Build GRCh38
rs945562538 Chr.10: 51698245-51698245 on Build GRCh38
rs758199123 Chr.10: 51698395-51698395 on Build GRCh38
rs374193081 Chr.10: 51698469-51698469 on Build GRCh38
rs772996942 Chr.10: 51698324-51698324 on Build GRCh38
rs755122614 Chr.10: 51698278-51698278 on Build GRCh38
rs778120310 Chr.10: 51698375-51698375 on Build GRCh38
rs774806602 Chr.10: 51698464-51698464 on Build GRCh38
rs768689903 Chr.10: 51698419-51698419 on Build GRCh38
rs1008374857 Chr.10: 51698303-51698303 on Build GRCh38
rs766792935 Chr.10: 51698338-51698338 on Build GRCh38
rs144891671 Chr.10: 51698289-51698289 on Build GRCh38
rs376951469 Chr.10: 51698348-51698348 on Build GRCh38
rs373436302 Chr.10: 51698433-51698433 on Build GRCh38
rs781265932 Chr.10: 51698279-51698279 on Build GRCh38
rs75877881 Chr.10: 51698437-51698437 on Build GRCh38
rs775981203 Chr.10: 51698294-51698294 on Build GRCh38
rs775371688 Chr.10: 51698254-51698254 on Build GRCh38
rs111331961 Chr.10: 51698273-51698273 on Build GRCh38
rs369368999 Chr.10: 51698283-51698283 on Build GRCh38
rs866839962 Chr.10: 51698466-51698466 on Build GRCh38
rs761210544 Chr.10: 51698319-51698319 on Build GRCh38
rs755471672 Chr.10: 51698353-51698353 on Build GRCh38
rs530966056 Chr.10: 51698259-51698259 on Build GRCh38
rs571544210 Chr.10: 51698257-51698257 on Build GRCh38
rs763471233 Chr.10: 51698331-51698331 on Build GRCh38
rs148622678 Chr.10: 51698300-51698300 on Build GRCh38
rs371185885 Chr.10: 51698362-51698362 on Build GRCh38
rs752100435 Chr.10: 51698277-51698277 on Build GRCh38
rs760341333 Chr.10: 51698322-51698322 on Build GRCh38
rs139186594 Chr.10: 51698286-51698286 on Build GRCh38
rs535610984 Chr.10: 51698400-51698400 on Build GRCh38
rs766879975 Chr.10: 51698454-51698454 on Build GRCh38
rs776404351 Chr.10: 51698426-51698426 on Build GRCh38
rs761490173 Chr.10: 51698261-51698261 on Build GRCh38
rs959432517 Chr.10: 51698380-51698380 on Build GRCh38
rs752868245 Chr.10: 51698366-51698366 on Build GRCh38

View Full Product Details