Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52239026-52239251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE9
Entrez Gene ID
9372
Gene Name
zinc finger FYVE-type containing 9
Gene Aliases
MADHIP, NSP, PPP1R173, SARA, SMADIP
UniGene
Hs.532345
Gene Chromosome Location
Chr.1: 52142001-52348664 on Build GRCh38
Transcripts
NM_004799.3 NM_007324.3 XM_011542437.2 XM_017002845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827052 c.1813G>A p.A605T Chr.1: 52239230-52239230 on Build GRCh38
414862 c.1787C>T p.S596L Chr.1: 52239204-52239204 on Build GRCh38
1343355 c.1827T>C p.N609N Chr.1: 52239244-52239244 on Build GRCh38
4742763 c.1613C>T p.T538I Chr.1: 52239030-52239030 on Build GRCh38
3360952 c.1750C>T p.P584S Chr.1: 52239167-52239167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000419162 Chr.1: 52239103-52239103 on Build GRCh38
rs757129930 Chr.1: 52239102-52239102 on Build GRCh38
rs780383286 Chr.1: 52239112-52239112 on Build GRCh38
rs536781182 Chr.1: 52239108-52239108 on Build GRCh38
rs764903872 Chr.1: 52239197-52239197 on Build GRCh38
rs758154233 Chr.1: 52239067-52239067 on Build GRCh38
rs373669433 Chr.1: 52239097-52239097 on Build GRCh38
rs776214144 Chr.1: 52239191-52239191 on Build GRCh38
rs201890566 Chr.1: 52239113-52239113 on Build GRCh38
rs368120815 Chr.1: 52239166-52239166 on Build GRCh38
rs145100186 Chr.1: 52239068-52239068 on Build GRCh38
rs751567594 Chr.1: 52239096-52239096 on Build GRCh38
rs149073048 Chr.1: 52239104-52239104 on Build GRCh38
rs370570538 Chr.1: 52239193-52239193 on Build GRCh38
rs147788994 Chr.1: 52239117-52239117 on Build GRCh38
rs370567015 Chr.1: 52239060-52239060 on Build GRCh38
rs3991041 Chr.1: 52239059-52239060 on Build GRCh38
rs112779296 Chr.1: 52239088-52239088 on Build GRCh38
rs762574495 Chr.1: 52239202-52239202 on Build GRCh38
rs774564734 Chr.1: 52239129-52239129 on Build GRCh38
rs764809480 Chr.1: 52239056-52239056 on Build GRCh38
rs772389529 Chr.1: 52239176-52239176 on Build GRCh38
rs574593980 Chr.1: 52239199-52239199 on Build GRCh38
rs997245541 Chr.1: 52239214-52239214 on Build GRCh38
rs769191238 Chr.1: 52239124-52239124 on Build GRCh38

View Full Product Details