Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56622612-56622839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF71
Entrez Gene ID
58491
Gene Name
zinc finger protein 71
Gene Aliases
EZFIT
UniGene
Hs.301431
Gene Chromosome Location
Chr.19: 56595259-56624481 on Build GRCh38
Transcripts
NM_021216.4 XM_011527193.2 XM_017027078.1 XM_017027079.1 XM_017027080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1271737 c.1413C>T p.C471C Chr.19: 56622700-56622700 on Build GRCh38
4466244 c.1422C>T p.T474T Chr.19: 56622709-56622709 on Build GRCh38
5519722 c.1372C>T p.R458W Chr.19: 56622659-56622659 on Build GRCh38
3540081 c.1344C>T p.I448I Chr.19: 56622631-56622631 on Build GRCh38
356890 c.1430G>A p.R477H Chr.19: 56622717-56622717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894542928 Chr.19: 56622772-56622772 on Build GRCh38
rs909041388 Chr.19: 56622702-56622702 on Build GRCh38
rs776488975 Chr.19: 56622796-56622796 on Build GRCh38
rs779577582 Chr.19: 56622670-56622670 on Build GRCh38
rs989269001 Chr.19: 56622634-56622634 on Build GRCh38
rs761769452 Chr.19: 56622656-56622656 on Build GRCh38
rs770985877 Chr.19: 56622644-56622644 on Build GRCh38
rs773773677 Chr.19: 56622643-56622643 on Build GRCh38
rs754511389 Chr.19: 56622746-56622746 on Build GRCh38
rs1046159320 Chr.19: 56622793-56622793 on Build GRCh38
rs141286207 Chr.19: 56622759-56622759 on Build GRCh38
rs568975591 Chr.19: 56622766-56622766 on Build GRCh38
rs767552340 Chr.19: 56622664-56622664 on Build GRCh38
rs145277321 Chr.19: 56622686-56622686 on Build GRCh38
rs772535342 Chr.19: 56622639-56622639 on Build GRCh38
rs200278100 Chr.19: 56622727-56622727 on Build GRCh38
rs765397884 Chr.19: 56622651-56622651 on Build GRCh38
rs373731127 Chr.19: 56622758-56622758 on Build GRCh38
rs142952966 Chr.19: 56622733-56622733 on Build GRCh38
rs771369788 Chr.19: 56622713-56622713 on Build GRCh38
rs942958411 Chr.19: 56622798-56622798 on Build GRCh38
rs369701185 Chr.19: 56622785-56622785 on Build GRCh38
rs775720585 Chr.19: 56622784-56622784 on Build GRCh38
rs569502405 Chr.19: 56622742-56622742 on Build GRCh38
rs759761031 Chr.19: 56622650-56622650 on Build GRCh38
rs745981856 Chr.19: 56622724-56622724 on Build GRCh38
rs201319397 Chr.19: 56622797-56622797 on Build GRCh38
rs910966492 Chr.19: 56622648-56622648 on Build GRCh38
rs750332150 Chr.19: 56622667-56622667 on Build GRCh38
rs776573515 Chr.19: 56622649-56622649 on Build GRCh38
rs778067986 Chr.19: 56622762-56622762 on Build GRCh38
rs778123469 Chr.19: 56622710-56622710 on Build GRCh38
rs758050277 Chr.19: 56622760-56622760 on Build GRCh38
rs773172735 Chr.19: 56622734-56622734 on Build GRCh38
rs772482422 Chr.19: 56622707-56622707 on Build GRCh38
rs745950321 Chr.19: 56622776-56622776 on Build GRCh38
rs763304877 Chr.19: 56622732-56622732 on Build GRCh38
rs766334335 Chr.19: 56622790-56622790 on Build GRCh38
rs770053922 Chr.19: 56622726-56622726 on Build GRCh38
rs548368256 Chr.19: 56622709-56622709 on Build GRCh38
rs760428575 Chr.19: 56622789-56622789 on Build GRCh38
rs776719783 Chr.19: 56622716-56622716 on Build GRCh38
rs769940962 Chr.19: 56622780-56622780 on Build GRCh38
rs764660464 Chr.19: 56622800-56622800 on Build GRCh38
rs773038921 Chr.19: 56622788-56622788 on Build GRCh38
rs748505911 Chr.19: 56622701-56622701 on Build GRCh38
rs747479050 Chr.19: 56622711-56622711 on Build GRCh38
rs753360449 Chr.19: 56622744-56622744 on Build GRCh38
rs766278120 Chr.19: 56622743-56622743 on Build GRCh38
rs754639538 Chr.19: 56622694-56622694 on Build GRCh38
rs537900353 Chr.19: 56622815-56622815 on Build GRCh38
rs976843416 Chr.19: 56622661-56622661 on Build GRCh38
rs757757496 Chr.19: 56622769-56622769 on Build GRCh38
rs748562974 Chr.19: 56622636-56622636 on Build GRCh38
rs778574971 Chr.19: 56622700-56622700 on Build GRCh38
rs969317484 Chr.19: 56622673-56622673 on Build GRCh38

View Full Product Details