Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12649010-12649245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
288885 c.2388C>T p.R796R Chr.19: 12649181-12649181 on Build GRCh38
1286072 c.2395G>T p.G799W Chr.19: 12649174-12649174 on Build GRCh38
4074380 c.2359A>T p.N787Y Chr.19: 12649210-12649210 on Build GRCh38
5916742 c.2353-4C>T p.? Chr.19: 12649220-12649220 on Build GRCh38
6706672 c.2386C>T p.R796C Chr.19: 12649183-12649183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367852398 Chr.19: 12649141-12649141 on Build GRCh38
rs745365527 Chr.19: 12649189-12649189 on Build GRCh38
rs952931725 Chr.19: 12649080-12649080 on Build GRCh38
rs148420017 Chr.19: 12649119-12649119 on Build GRCh38
rs544562918 Chr.19: 12649075-12649075 on Build GRCh38
rs770430333 Chr.19: 12649091-12649091 on Build GRCh38
rs864621994 Chr.19: 12649170-12649170 on Build GRCh38
rs762300861 Chr.19: 12649198-12649198 on Build GRCh38
rs762235411 Chr.19: 12649137-12649137 on Build GRCh38
rs964362143 Chr.19: 12649057-12649057 on Build GRCh38
rs955076933 Chr.19: 12649211-12649211 on Build GRCh38
rs992957181 Chr.19: 12649150-12649150 on Build GRCh38
rs141391488 Chr.19: 12649168-12649168 on Build GRCh38
rs147850230 Chr.19: 12649163-12649163 on Build GRCh38
rs777432395 Chr.19: 12649178-12649178 on Build GRCh38
rs1008690907 Chr.19: 12649033-12649033 on Build GRCh38
rs975398739 Chr.19: 12649062-12649062 on Build GRCh38
rs748767803 Chr.19: 12649087-12649087 on Build GRCh38
rs777253267 Chr.19: 12649195-12649195 on Build GRCh38
rs772831871 Chr.19: 12649086-12649086 on Build GRCh38
rs750957781 Chr.19: 12649182-12649182 on Build GRCh38
rs748899790 Chr.19: 12649181-12649181 on Build GRCh38
rs778543070 Chr.19: 12649188-12649188 on Build GRCh38
rs797044680 Chr.19: 12649169-12649170 on Build GRCh38
rs775862875 Chr.19: 12649093-12649093 on Build GRCh38
rs772742355 Chr.19: 12649183-12649183 on Build GRCh38
rs892632100 Chr.19: 12649127-12649127 on Build GRCh38
rs142702682 Chr.19: 12649171-12649171 on Build GRCh38
rs756302695 Chr.19: 12649055-12649055 on Build GRCh38
rs920643986 Chr.19: 12649053-12649053 on Build GRCh38
rs398123456 Chr.19: 12649174-12649174 on Build GRCh38
rs769324462 Chr.19: 12649192-12649192 on Build GRCh38
rs398123457 Chr.19: 12649134-12649134 on Build GRCh38
rs948132304 Chr.19: 12649054-12649054 on Build GRCh38
rs762947868 Chr.19: 12649148-12649148 on Build GRCh38
rs764685302 Chr.19: 12649110-12649110 on Build GRCh38
rs200036864 Chr.19: 12649135-12649135 on Build GRCh38
rs80338681 Chr.19: 12649146-12649146 on Build GRCh38
rs780214785 Chr.19: 12649199-12649199 on Build GRCh38

View Full Product Details