Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161117928-161118199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001185092.1 NM_001185093.1 NM_001185094.1 NM_005600.2 XM_005245214.2 XM_005245215.4 XM_005245216.3 XM_006711341.3 XM_017001371.1 NM_012394.3 XM_011509624.2

Gene Symbol
PFDN2
Entrez Gene ID
5202
Gene Name
prefoldin subunit 2
Gene Aliases
PFD2
UniGene
Hs.492516
Gene Chromosome Location
Chr.1: 161100556-161118076 on Build GRCh38
Transcripts
NM_001185092.1 NM_001185093.1 NM_001185094.1 NM_005600.2 XM_005245214.2 XM_005245215.4 XM_005245216.3 XM_006711341.3 XM_017001371.1 NM_012394.3 XM_011509624.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5670151 c.2+3A>G p.? Chr.1: 161118181-161118181 on Build GRCh38
333616 c.48G>T p.A16A Chr.1: 161117979-161117979 on Build GRCh38
4432018 c.12C>G p.N4K Chr.1: 161118015-161118015 on Build GRCh38
5566867 c.19C>G p.R7G Chr.1: 161118008-161118008 on Build GRCh38
1261970 c.22G>T p.A8S Chr.1: 161118005-161118005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917152727 Chr.1: 161118089-161118089 on Build GRCh38
rs987175033 Chr.1: 161118043-161118043 on Build GRCh38
rs754144546 Chr.1: 161118140-161118140 on Build GRCh38
rs745796523 Chr.1: 161118056-161118056 on Build GRCh38
rs146025814 Chr.1: 161118003-161118003 on Build GRCh38
rs756473177 Chr.1: 161118027-161118027 on Build GRCh38
rs537093405 Chr.1: 161118118-161118118 on Build GRCh38
rs746514957 Chr.1: 161118128-161118128 on Build GRCh38
rs891391978 Chr.1: 161118142-161118142 on Build GRCh38
rs767248289 Chr.1: 161117951-161117951 on Build GRCh38
rs1047070177 Chr.1: 161118099-161118099 on Build GRCh38
rs978424102 Chr.1: 161118042-161118042 on Build GRCh38
rs764458045 Chr.1: 161118088-161118088 on Build GRCh38
rs571215666 Chr.1: 161118106-161118106 on Build GRCh38
rs767508343 Chr.1: 161118019-161118019 on Build GRCh38
rs778112334 Chr.1: 161118028-161118028 on Build GRCh38
rs377135940 Chr.1: 161118133-161118133 on Build GRCh38
rs752994151 Chr.1: 161118065-161118065 on Build GRCh38
rs34402935 Chr.1: 161118090-161118091 on Build GRCh38
rs200572879 Chr.1: 161118145-161118145 on Build GRCh38
rs967333892 Chr.1: 161118030-161118030 on Build GRCh38
rs760330512 Chr.1: 161118007-161118007 on Build GRCh38
rs937150559 Chr.1: 161118086-161118086 on Build GRCh38
rs752313527 Chr.1: 161117960-161117960 on Build GRCh38
rs967163093 Chr.1: 161118055-161118055 on Build GRCh38
rs377183729 Chr.1: 161118130-161118130 on Build GRCh38
rs551733325 Chr.1: 161118124-161118124 on Build GRCh38
rs770980275 Chr.1: 161117945-161117945 on Build GRCh38
rs774916342 Chr.1: 161118127-161118127 on Build GRCh38
rs752865281 Chr.1: 161118135-161118135 on Build GRCh38
rs369706655 Chr.1: 161118169-161118169 on Build GRCh38
rs765587886 Chr.1: 161117982-161117982 on Build GRCh38
rs770084985 Chr.1: 161117994-161117994 on Build GRCh38
rs757433697 Chr.1: 161118144-161118144 on Build GRCh38
rs1038412831 Chr.1: 161118109-161118109 on Build GRCh38
rs949983882 Chr.1: 161118098-161118098 on Build GRCh38
rs1032748864 Chr.1: 161118157-161118157 on Build GRCh38
rs375173230 Chr.1: 161117985-161117985 on Build GRCh38
rs1017064640 Chr.1: 161118005-161118005 on Build GRCh38
rs780618093 Chr.1: 161118151-161118151 on Build GRCh38
rs777770359 Chr.1: 161117997-161117997 on Build GRCh38
rs375266818 Chr.1: 161118012-161118012 on Build GRCh38
rs958550998 Chr.1: 161118161-161118161 on Build GRCh38
rs756971354 Chr.1: 161118139-161118139 on Build GRCh38
rs899970421 Chr.1: 161118006-161118006 on Build GRCh38
rs778729243 Chr.1: 161118038-161118038 on Build GRCh38
rs752041833 Chr.1: 161118018-161118018 on Build GRCh38
rs557429482 Chr.1: 161118082-161118082 on Build GRCh38
rs758959211 Chr.1: 161117984-161117984 on Build GRCh38
rs766720552 Chr.1: 161118014-161118014 on Build GRCh38
rs376601895 Chr.1: 161118037-161118037 on Build GRCh38
rs776914738 Chr.1: 161118108-161118108 on Build GRCh38
rs370857008 Chr.1: 161118137-161118137 on Build GRCh38
rs557271646 Chr.1: 161118136-161118136 on Build GRCh38
rs200205031 Chr.1: 161118173-161118173 on Build GRCh38
rs553155473 Chr.1: 161118154-161118154 on Build GRCh38
rs530213094 Chr.1: 161117946-161117946 on Build GRCh38
rs967446824 Chr.1: 161118150-161118150 on Build GRCh38
rs761419606 Chr.1: 161118131-161118131 on Build GRCh38
rs771031486 Chr.1: 161118000-161118000 on Build GRCh38
rs774646286 Chr.1: 161118158-161118158 on Build GRCh38
rs546804573 Chr.1: 161117998-161117998 on Build GRCh38
rs142691653 Chr.1: 161117990-161117990 on Build GRCh38
rs900042545 Chr.1: 161118114-161118114 on Build GRCh38
rs774132214 Chr.1: 161118002-161118002 on Build GRCh38
rs1020343032 Chr.1: 161118053-161118053 on Build GRCh38
rs767939258 Chr.1: 161118064-161118065 on Build GRCh38
rs749143314 Chr.1: 161118033-161118033 on Build GRCh38
rs771983660 Chr.1: 161118071-161118071 on Build GRCh38
rs185780451 Chr.1: 161118149-161118149 on Build GRCh38
rs758331343 Chr.1: 161118146-161118146 on Build GRCh38
rs573858934 Chr.1: 161118021-161118021 on Build GRCh38
rs781295624 Chr.1: 161117993-161117993 on Build GRCh38
rs762160518 Chr.1: 161118112-161118112 on Build GRCh38
rs760279623 Chr.1: 161117976-161117976 on Build GRCh38
rs780007246 Chr.1: 161118147-161118147 on Build GRCh38
rs950067642 Chr.1: 161117968-161117968 on Build GRCh38
rs565609283 Chr.1: 161118008-161118008 on Build GRCh38
rs772505496 Chr.1: 161118172-161118172 on Build GRCh38
rs1020349074 Chr.1: 161118165-161118165 on Build GRCh38
rs750724780 Chr.1: 161117983-161117983 on Build GRCh38
rs768066275 Chr.1: 161118134-161118134 on Build GRCh38
rs766661058 Chr.1: 161118132-161118132 on Build GRCh38
rs773630514 Chr.1: 161118011-161118011 on Build GRCh38

View Full Product Details