Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104087823-104088080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCFC2
Entrez Gene ID
29915
Gene Name
host cell factor C2
Gene Aliases
HCF-2, HCF2
UniGene
Hs.506558
Gene Chromosome Location
Chr.12: 104064458-104108954 on Build GRCh38
Transcripts
NM_013320.2 XM_017019241.1 XM_017019242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934225 c.1284+2T>C p.? Chr.12: 104088040-104088040 on Build GRCh38
5793246 c.1283G>A p.S428N Chr.12: 104088037-104088037 on Build GRCh38
691514 c.1283G>C p.S428T Chr.12: 104088037-104088037 on Build GRCh38
6441886 c.1232-1G>A p.? Chr.12: 104087985-104087985 on Build GRCh38
5587963 c.1272C>T p.I424I Chr.12: 104088026-104088026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1047958924 Chr.12: 104087854-104087854 on Build GRCh38
rs1001952893 Chr.12: 104087914-104087914 on Build GRCh38
rs1032209183 Chr.12: 104087932-104087932 on Build GRCh38
rs542271303 Chr.12: 104088020-104088020 on Build GRCh38
rs1020845522 Chr.12: 104088013-104088013 on Build GRCh38
rs140444104 Chr.12: 104088027-104088027 on Build GRCh38
rs537847117 Chr.12: 104087864-104087864 on Build GRCh38
rs749830651 Chr.12: 104087969-104087969 on Build GRCh38
rs373952165 Chr.12: 104088015-104088015 on Build GRCh38
rs776239739 Chr.12: 104087936-104087936 on Build GRCh38
rs542872865 Chr.12: 104087975-104087975 on Build GRCh38
rs112756196 Chr.12: 104087978-104087978 on Build GRCh38
rs1024739020 Chr.12: 104087909-104087909 on Build GRCh38
rs886793562 Chr.12: 104087865-104087865 on Build GRCh38
rs34811234 Chr.12: 104087913-104087913 on Build GRCh38
rs761647344 Chr.12: 104087954-104087954 on Build GRCh38
rs957881007 Chr.12: 104087958-104087958 on Build GRCh38
rs930660652 Chr.12: 104087955-104087955 on Build GRCh38
rs76655421 Chr.12: 104087902-104087902 on Build GRCh38
rs151222227 Chr.12: 104088016-104088016 on Build GRCh38

View Full Product Details