Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12029734-12029969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIIP
Entrez Gene ID
60672
Gene Name
migration and invasion inhibitory protein
Gene Aliases
IIP45
UniGene
Hs.8595
Gene Chromosome Location
Chr.1: 12019242-12032049 on Build GRCh38
Transcripts
NM_021933.3 XM_005263487.3 XM_011541895.1 XM_011541896.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6712790 c.718G>A p.V240M Chr.1: 12029767-12029767 on Build GRCh38
5549355 c.828G>A p.A276A Chr.1: 12029877-12029877 on Build GRCh38
2180249 c.731G>A p.R244H Chr.1: 12029780-12029780 on Build GRCh38
6057770 c.741G>T p.R247R Chr.1: 12029790-12029790 on Build GRCh38
3472352 c.767C>T p.P256L Chr.1: 12029816-12029816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747127837 Chr.1: 12029881-12029881 on Build GRCh38
rs999586778 Chr.1: 12029819-12029819 on Build GRCh38
rs373610599 Chr.1: 12029903-12029903 on Build GRCh38
rs776402361 Chr.1: 12029800-12029812 on Build GRCh38
rs755823829 Chr.1: 12029860-12029860 on Build GRCh38
rs902612989 Chr.1: 12029818-12029818 on Build GRCh38
rs200661070 Chr.1: 12029907-12029907 on Build GRCh38
rs777956456 Chr.1: 12029825-12029825 on Build GRCh38
rs748247221 Chr.1: 12029781-12029781 on Build GRCh38
rs771058460 Chr.1: 12029885-12029885 on Build GRCh38
rs780949591 Chr.1: 12029812-12029812 on Build GRCh38
rs770951582 Chr.1: 12029791-12029792 on Build GRCh38
rs201594506 Chr.1: 12029890-12029890 on Build GRCh38
rs377345297 Chr.1: 12029917-12029917 on Build GRCh38
rs769648587 Chr.1: 12029840-12029840 on Build GRCh38
rs370000251 Chr.1: 12029877-12029877 on Build GRCh38
rs139665248 Chr.1: 12029835-12029835 on Build GRCh38
rs763703048 Chr.1: 12029798-12029798 on Build GRCh38
rs1139720 Chr.1: 12029848-12029848 on Build GRCh38
rs112758519 Chr.1: 12029916-12029916 on Build GRCh38
rs753111504 Chr.1: 12029941-12029941 on Build GRCh38
rs35720158 Chr.1: 12029846-12029846 on Build GRCh38
rs1024110747 Chr.1: 12029839-12029839 on Build GRCh38
rs759440745 Chr.1: 12029921-12029921 on Build GRCh38
rs781245805 Chr.1: 12029886-12029886 on Build GRCh38
rs760452156 Chr.1: 12029921-12029921 on Build GRCh38
rs142095617 Chr.1: 12029801-12029801 on Build GRCh38
rs143363945 Chr.1: 12029796-12029796 on Build GRCh38
rs2295289 Chr.1: 12029788-12029788 on Build GRCh38
rs779460357 Chr.1: 12029861-12029861 on Build GRCh38
rs41278640 Chr.1: 12029767-12029767 on Build GRCh38
rs756959119 Chr.1: 12029802-12029802 on Build GRCh38
rs996184721 Chr.1: 12029845-12029845 on Build GRCh38
rs374656152 Chr.1: 12029789-12029789 on Build GRCh38
rs367745113 Chr.1: 12029791-12029791 on Build GRCh38
rs749618686 Chr.1: 12029778-12029778 on Build GRCh38
rs949302780 Chr.1: 12029927-12029927 on Build GRCh38
rs1032346352 Chr.1: 12029833-12029833 on Build GRCh38
rs117146839 Chr.1: 12029889-12029889 on Build GRCh38
rs755333128 Chr.1: 12029815-12029815 on Build GRCh38
rs759261675 Chr.1: 12029792-12029792 on Build GRCh38
rs112218993 Chr.1: 12029779-12029779 on Build GRCh38
rs375352476 Chr.1: 12029853-12029853 on Build GRCh38
rs922131860 Chr.1: 12029761-12029761 on Build GRCh38
rs756831588 Chr.1: 12029766-12029766 on Build GRCh38
rs754256847 Chr.1: 12029813-12029813 on Build GRCh38
rs766077696 Chr.1: 12029937-12029937 on Build GRCh38
rs758965693 Chr.1: 12029876-12029876 on Build GRCh38
rs748731371 Chr.1: 12029821-12029821 on Build GRCh38
rs769985165 Chr.1: 12029888-12029888 on Build GRCh38
rs80150922 Chr.1: 12029817-12029817 on Build GRCh38
rs777138221 Chr.1: 12029757-12029757 on Build GRCh38
rs747011999 Chr.1: 12029792-12029804 on Build GRCh38
rs752688921 Chr.1: 12029795-12029795 on Build GRCh38
rs1013859842 Chr.1: 12029816-12029816 on Build GRCh38
rs372139970 Chr.1: 12029830-12029830 on Build GRCh38
rs2295290 Chr.1: 12029794-12029794 on Build GRCh38
rs772251426 Chr.1: 12029786-12029786 on Build GRCh38
rs761157763 Chr.1: 12029849-12029849 on Build GRCh38
rs746132806 Chr.1: 12029887-12029887 on Build GRCh38
rs775238102 Chr.1: 12029841-12029841 on Build GRCh38
rs958294515 Chr.1: 12029756-12029756 on Build GRCh38
rs367987372 Chr.1: 12029760-12029760 on Build GRCh38
rs750154144 Chr.1: 12029858-12029858 on Build GRCh38
rs752980208 Chr.1: 12029871-12029871 on Build GRCh38
rs373485692 Chr.1: 12029831-12029831 on Build GRCh38
rs4514248 Chr.1: 12029944-12029944 on Build GRCh38
rs772426039 Chr.1: 12029823-12029823 on Build GRCh38
rs146566756 Chr.1: 12029780-12029780 on Build GRCh38
rs778030939 Chr.1: 12029759-12029774 on Build GRCh38
rs763891053 Chr.1: 12029847-12029847 on Build GRCh38
rs768640719 Chr.1: 12029896-12029896 on Build GRCh38
rs138345116 Chr.1: 12029790-12029790 on Build GRCh38

View Full Product Details