Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74663856-74664107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AREL1
Entrez Gene ID
9870
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Gene Aliases
FIEL1, KIAA0317
UniGene
Hs.497417
Gene Chromosome Location
Chr.14: 74661252-74713118 on Build GRCh38
Transcripts
NM_001039479.1 XM_005268244.3 XM_006720344.2 XM_011537415.2 XM_011537416.2 XM_011537417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7139829 c.2291G>C p.G764A Chr.14: 74663977-74663977 on Build GRCh38
138053 c.2236G>A p.E746K Chr.14: 74664032-74664032 on Build GRCh38
5812480 c.2369+3A>G p.? Chr.14: 74663896-74663896 on Build GRCh38
6141278 c.2348C>T p.T783M Chr.14: 74663920-74663920 on Build GRCh38
5648470 c.2200A>T p.R734W Chr.14: 74664068-74664068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780533710 Chr.14: 74664009-74664009 on Build GRCh38
rs151053775 Chr.14: 74663955-74663955 on Build GRCh38
rs780090234 Chr.14: 74663892-74663892 on Build GRCh38
rs985472679 Chr.14: 74663877-74663877 on Build GRCh38
rs778679369 Chr.14: 74664027-74664027 on Build GRCh38
rs1050446804 Chr.14: 74663928-74663928 on Build GRCh38
rs201605025 Chr.14: 74664017-74664017 on Build GRCh38
rs112083260 Chr.14: 74663937-74663937 on Build GRCh38
rs772203633 Chr.14: 74664054-74664054 on Build GRCh38
rs756890860 Chr.14: 74663979-74663979 on Build GRCh38
rs768507976 Chr.14: 74663919-74663919 on Build GRCh38
rs762039755 Chr.14: 74663920-74663920 on Build GRCh38
rs374039077 Chr.14: 74663994-74663994 on Build GRCh38
rs773529888 Chr.14: 74664066-74664066 on Build GRCh38
rs373837829 Chr.14: 74663905-74663905 on Build GRCh38
rs748325871 Chr.14: 74664042-74664042 on Build GRCh38
rs370313422 Chr.14: 74663966-74663966 on Build GRCh38
rs371610162 Chr.14: 74663932-74663932 on Build GRCh38
rs535117829 Chr.14: 74663967-74663967 on Build GRCh38
rs141315233 Chr.14: 74664015-74664015 on Build GRCh38
rs1040013789 Chr.14: 74664026-74664026 on Build GRCh38
rs979242173 Chr.14: 74663933-74663933 on Build GRCh38
rs376915333 Chr.14: 74663931-74663931 on Build GRCh38
rs759974019 Chr.14: 74663964-74663964 on Build GRCh38
rs1040357693 Chr.14: 74663901-74663901 on Build GRCh38
rs569604261 Chr.14: 74663958-74663958 on Build GRCh38
rs369603208 Chr.14: 74663884-74663884 on Build GRCh38
rs777263416 Chr.14: 74664074-74664074 on Build GRCh38
rs184450822 Chr.14: 74663885-74663885 on Build GRCh38
rs189402492 Chr.14: 74664019-74664019 on Build GRCh38
rs769643137 Chr.14: 74663887-74663887 on Build GRCh38
rs200776635 Chr.14: 74664071-74664071 on Build GRCh38
rs765657877 Chr.14: 74664088-74664088 on Build GRCh38
rs760133498 Chr.14: 74664085-74664085 on Build GRCh38
rs781111595 Chr.14: 74663885-74663885 on Build GRCh38
rs767005007 Chr.14: 74663981-74663981 on Build GRCh38
rs749285374 Chr.14: 74664020-74664020 on Build GRCh38
rs754713908 Chr.14: 74664021-74664021 on Build GRCh38
rs372593008 Chr.14: 74663936-74663936 on Build GRCh38

View Full Product Details