Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38222310-38222577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1755
Entrez Gene ID
85449
Gene Name
KIAA1755
Gene Aliases
-
UniGene
Hs.472690
Gene Chromosome Location
Chr.20: 38210505-38260772 on Build GRCh38
Transcripts
NM_001029864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243737 c.2328C>G p.P776P Chr.20: 38222538-38222538 on Build GRCh38
5631894 c.2299G>A p.E767K Chr.20: 38222567-38222567 on Build GRCh38
4695788 c.2347C>T p.R783W Chr.20: 38222519-38222519 on Build GRCh38
4581784 c.2364C>G p.T788T Chr.20: 38222502-38222502 on Build GRCh38
723772 c.2410G>A p.D804N Chr.20: 38222456-38222456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768096004 Chr.20: 38222417-38222417 on Build GRCh38
rs565370903 Chr.20: 38222405-38222405 on Build GRCh38
rs984062968 Chr.20: 38222503-38222503 on Build GRCh38
rs923540411 Chr.20: 38222478-38222478 on Build GRCh38
rs866928531 Chr.20: 38222390-38222390 on Build GRCh38
rs780692847 Chr.20: 38222448-38222448 on Build GRCh38
rs565906736 Chr.20: 38222451-38222451 on Build GRCh38
rs747693591 Chr.20: 38222537-38222537 on Build GRCh38
rs144432535 Chr.20: 38222333-38222333 on Build GRCh38
rs142952405 Chr.20: 38222372-38222372 on Build GRCh38
rs767518364 Chr.20: 38222505-38222505 on Build GRCh38
rs770635860 Chr.20: 38222545-38222545 on Build GRCh38
rs190340907 Chr.20: 38222536-38222536 on Build GRCh38
rs762784986 Chr.20: 38222477-38222477 on Build GRCh38
rs774220205 Chr.20: 38222501-38222501 on Build GRCh38
rs1025834429 Chr.20: 38222375-38222376 on Build GRCh38
rs746383757 Chr.20: 38222440-38222440 on Build GRCh38
rs536659875 Chr.20: 38222502-38222502 on Build GRCh38
rs201309851 Chr.20: 38222507-38222507 on Build GRCh38
rs772819694 Chr.20: 38222475-38222475 on Build GRCh38
rs367603291 Chr.20: 38222433-38222433 on Build GRCh38
rs764464550 Chr.20: 38222509-38222509 on Build GRCh38
rs201601539 Chr.20: 38222518-38222518 on Build GRCh38
rs375892439 Chr.20: 38222556-38222556 on Build GRCh38
rs970520160 Chr.20: 38222348-38222348 on Build GRCh38
rs569990901 Chr.20: 38222519-38222519 on Build GRCh38
rs952400654 Chr.20: 38222391-38222391 on Build GRCh38
rs757678414 Chr.20: 38222435-38222435 on Build GRCh38
rs200339630 Chr.20: 38222441-38222441 on Build GRCh38
rs199571448 Chr.20: 38222514-38222514 on Build GRCh38
rs374416714 Chr.20: 38222406-38222406 on Build GRCh38
rs796797536 Chr.20: 38222479-38222479 on Build GRCh38
rs760806149 Chr.20: 38222508-38222508 on Build GRCh38
rs531770107 Chr.20: 38222362-38222362 on Build GRCh38
rs748932889 Chr.20: 38222544-38222544 on Build GRCh38
rs999793944 Chr.20: 38222351-38222351 on Build GRCh38
rs150629642 Chr.20: 38222485-38222485 on Build GRCh38
rs769392337 Chr.20: 38222469-38222469 on Build GRCh38
rs779350424 Chr.20: 38222439-38222439 on Build GRCh38
rs974943586 Chr.20: 38222473-38222473 on Build GRCh38
rs41310010 Chr.20: 38222538-38222538 on Build GRCh38
rs561282999 Chr.20: 38222342-38222342 on Build GRCh38

View Full Product Details