Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41437884-41438155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT38
Entrez Gene ID
8687
Gene Name
keratin 38
Gene Aliases
HA8, KRTHA8, hHa8
UniGene
Hs.248188
Gene Chromosome Location
Chr.17: 41436369-41441344 on Build GRCh38
Transcripts
NM_006771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323990 c.1230C>A p.S410R Chr.17: 41438104-41438104 on Build GRCh38
1256108 c.1227A>C p.E409D Chr.17: 41438107-41438107 on Build GRCh38
3517367 c.1225G>A p.E409K Chr.17: 41438109-41438109 on Build GRCh38
5852501 c.1214G>A p.R405Q Chr.17: 41438120-41438120 on Build GRCh38
329690 c.1241+10delA p.? Chr.17: 41438083-41438083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs118129392 Chr.17: 41438020-41438020 on Build GRCh38
rs528854755 Chr.17: 41437919-41437919 on Build GRCh38
rs998082885 Chr.17: 41438103-41438103 on Build GRCh38
rs202116349 Chr.17: 41438084-41438084 on Build GRCh38
rs28550887 Chr.17: 41438104-41438104 on Build GRCh38
rs767358006 Chr.17: 41438126-41438126 on Build GRCh38
rs750341391 Chr.17: 41438132-41438132 on Build GRCh38
rs778151960 Chr.17: 41438088-41438088 on Build GRCh38
rs940192085 Chr.17: 41437946-41437946 on Build GRCh38
rs1056755511 Chr.17: 41437969-41437969 on Build GRCh38
rs909941094 Chr.17: 41437935-41437935 on Build GRCh38
rs998724980 Chr.17: 41438061-41438061 on Build GRCh38
rs200202096 Chr.17: 41438080-41438080 on Build GRCh38
rs534136162 Chr.17: 41438083-41438083 on Build GRCh38
rs762408014 Chr.17: 41438034-41438034 on Build GRCh38
rs1006715257 Chr.17: 41437983-41437983 on Build GRCh38
rs373583387 Chr.17: 41438058-41438058 on Build GRCh38
rs1052673386 Chr.17: 41437927-41437927 on Build GRCh38
rs758616083 Chr.17: 41438085-41438085 on Build GRCh38
rs752920640 Chr.17: 41438082-41438082 on Build GRCh38
rs768404914 Chr.17: 41438121-41438121 on Build GRCh38
rs965283869 Chr.17: 41437906-41437906 on Build GRCh38
rs774100660 Chr.17: 41438123-41438123 on Build GRCh38
rs761736391 Chr.17: 41438125-41438125 on Build GRCh38
rs745806840 Chr.17: 41437929-41437940 on Build GRCh38
rs72828202 Chr.17: 41438134-41438134 on Build GRCh38
rs1016302301 Chr.17: 41438010-41438010 on Build GRCh38
rs746326510 Chr.17: 41438100-41438100 on Build GRCh38
rs771242795 Chr.17: 41438089-41438089 on Build GRCh38
rs902843785 Chr.17: 41437913-41437913 on Build GRCh38
rs775058782 Chr.17: 41438116-41438116 on Build GRCh38
rs761893947 Chr.17: 41438043-41438043 on Build GRCh38
rs939600562 Chr.17: 41437959-41437959 on Build GRCh38
rs767587112 Chr.17: 41438046-41438046 on Build GRCh38
rs2071613 Chr.17: 41437952-41437952 on Build GRCh38
rs1028728355 Chr.17: 41437920-41437920 on Build GRCh38
rs997702114 Chr.17: 41437949-41437949 on Build GRCh38
rs4796789 Chr.17: 41437933-41437933 on Build GRCh38
rs953490812 Chr.17: 41437975-41437975 on Build GRCh38
rs972031721 Chr.17: 41438026-41438026 on Build GRCh38
rs755021748 Chr.17: 41438068-41438068 on Build GRCh38
rs762549714 Chr.17: 41438120-41438120 on Build GRCh38
rs376566657 Chr.17: 41438019-41438019 on Build GRCh38
rs758408839 Chr.17: 41437967-41437967 on Build GRCh38
rs539724527 Chr.17: 41437971-41437971 on Build GRCh38

View Full Product Details