Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39513262-39513535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOBP
Entrez Gene ID
4336
Gene Name
myelin-associated oligodendrocyte basic protein
Gene Aliases
-
UniGene
Hs.121333
Gene Chromosome Location
Chr.3: 39467573-39529497 on Build GRCh38
Transcripts
NM_001278322.1 NM_182935.3 NR_003090.2 NR_103504.1 NR_103505.1 NR_103506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1692623 c.232C>T p.P78S Chr.3: 39513408-39513408 on Build GRCh38
4938682 c.239A>G p.K80R Chr.3: 39513415-39513415 on Build GRCh38
2987670 c.237G>T p.K79N Chr.3: 39513413-39513413 on Build GRCh38
5399336 c.233C>G p.P78R Chr.3: 39513409-39513409 on Build GRCh38
3660546 c.233C>T p.P78L Chr.3: 39513409-39513409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377441394 Chr.3: 39513468-39513468 on Build GRCh38
rs767504231 Chr.3: 39513345-39513345 on Build GRCh38
rs769546332 Chr.3: 39513456-39513456 on Build GRCh38
rs188200484 Chr.3: 39513342-39513342 on Build GRCh38
rs779455244 Chr.3: 39513395-39513395 on Build GRCh38
rs553047971 Chr.3: 39513370-39513370 on Build GRCh38
rs372933674 Chr.3: 39513340-39513340 on Build GRCh38
rs550786049 Chr.3: 39513315-39513315 on Build GRCh38
rs771552516 Chr.3: 39513373-39513373 on Build GRCh38
rs900424906 Chr.3: 39513321-39513321 on Build GRCh38
rs374697287 Chr.3: 39513328-39513328 on Build GRCh38
rs754177769 Chr.3: 39513362-39513362 on Build GRCh38
rs889492858 Chr.3: 39513505-39513505 on Build GRCh38
rs1048544606 Chr.3: 39513460-39513460 on Build GRCh38
rs776474242 Chr.3: 39513442-39513442 on Build GRCh38
rs983582269 Chr.3: 39513465-39513465 on Build GRCh38
rs552724 Chr.3: 39513455-39513455 on Build GRCh38
rs2233204 Chr.3: 39513295-39513295 on Build GRCh38
rs762817109 Chr.3: 39513461-39513461 on Build GRCh38
rs752685514 Chr.3: 39513472-39513472 on Build GRCh38
rs778008267 Chr.3: 39513353-39513353 on Build GRCh38
rs1014687735 Chr.3: 39513483-39513483 on Build GRCh38
rs918517943 Chr.3: 39513419-39513419 on Build GRCh38
rs538695337 Chr.3: 39513489-39513489 on Build GRCh38
rs752911099 Chr.3: 39513352-39513352 on Build GRCh38
rs1008886338 Chr.3: 39513301-39513301 on Build GRCh38
rs746622008 Chr.3: 39513411-39513411 on Build GRCh38
rs757676152 Chr.3: 39513368-39513368 on Build GRCh38
rs534239256 Chr.3: 39513421-39513421 on Build GRCh38
rs757472283 Chr.3: 39513508-39513508 on Build GRCh38
rs537226501 Chr.3: 39513313-39513313 on Build GRCh38
rs369235076 Chr.3: 39513415-39513415 on Build GRCh38
rs779469322 Chr.3: 39513369-39513369 on Build GRCh38
rs773140830 Chr.3: 39513458-39513458 on Build GRCh38

View Full Product Details