Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 166849434-166849659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POGK
Entrez Gene ID
57645
Gene Name
pogo transposable element with KRAB domain
Gene Aliases
BASS2, KRBOX2, LST003
UniGene
Hs.432752
Gene Chromosome Location
Chr.1: 166839404-166856344 on Build GRCh38
Transcripts
NM_001314014.1 NM_017542.4 XM_005245370.3 XM_011509814.2 XM_011509815.1 XM_017001914.1 XM_017001915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6992320 c.1000G>C p.D334H Chr.1: 166849579-166849579 on Build GRCh38
2185294 c.1080G>C p.M360I Chr.1: 166849659-166849659 on Build GRCh38
414357 c.931C>G p.R311G Chr.1: 166849510-166849510 on Build GRCh38
4851245 c.985C>A p.Q329K Chr.1: 166849564-166849564 on Build GRCh38
898973 c.890A>G p.N297S Chr.1: 166849469-166849469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369140999 Chr.1: 166849575-166849575 on Build GRCh38
rs778117468 Chr.1: 166849482-166849482 on Build GRCh38
rs746220367 Chr.1: 166849629-166849629 on Build GRCh38
rs181432813 Chr.1: 166849611-166849611 on Build GRCh38
rs772385405 Chr.1: 166849634-166849634 on Build GRCh38
rs770387099 Chr.1: 166849477-166849477 on Build GRCh38
rs1022465439 Chr.1: 166849569-166849569 on Build GRCh38
rs759690920 Chr.1: 166849537-166849537 on Build GRCh38
rs753785988 Chr.1: 166849574-166849574 on Build GRCh38
rs775699186 Chr.1: 166849557-166849557 on Build GRCh38
rs371192826 Chr.1: 166849515-166849515 on Build GRCh38
rs41269682 Chr.1: 166849494-166849494 on Build GRCh38
rs374953651 Chr.1: 166849545-166849545 on Build GRCh38
rs760649233 Chr.1: 166849558-166849558 on Build GRCh38
rs750103246 Chr.1: 166849582-166849582 on Build GRCh38
rs143821995 Chr.1: 166849635-166849635 on Build GRCh38
rs778369717 Chr.1: 166849610-166849610 on Build GRCh38
rs890794290 Chr.1: 166849584-166849584 on Build GRCh38
rs1011963569 Chr.1: 166849533-166849533 on Build GRCh38
rs148246779 Chr.1: 166849597-166849597 on Build GRCh38
rs774571825 Chr.1: 166849520-166849520 on Build GRCh38
rs764133929 Chr.1: 166849563-166849563 on Build GRCh38
rs760624347 Chr.1: 166849620-166849620 on Build GRCh38
rs758133548 Chr.1: 166849594-166849594 on Build GRCh38
rs970273709 Chr.1: 166849626-166849626 on Build GRCh38
rs746494790 Chr.1: 166849455-166849455 on Build GRCh38
rs143063474 Chr.1: 166849602-166849602 on Build GRCh38
rs771291864 Chr.1: 166849617-166849617 on Build GRCh38

View Full Product Details