Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 1611622-1611871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXC1
Entrez Gene ID
2296
Gene Name
forkhead box C1
Gene Aliases
ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3
UniGene
Hs.348883
Gene Chromosome Location
Chr.6: 1610446-1613897 on Build GRCh38
Transcripts
NM_001453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382109 c.1250G>C p.G417A Chr.6: 1611695-1611695 on Build GRCh38
6547052 c.1305G>T p.P435P Chr.6: 1611750-1611750 on Build GRCh38
5030728 c.1339_1350del12 p.G453_G456delGGGG Chr.6: 1611784-1611795 on Build GRCh38
6382723 c.1249G>C p.G417R Chr.6: 1611694-1611694 on Build GRCh38
6513715 c.1415C>A p.S472* Chr.6: 1611860-1611860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373075286 Chr.6: 1611787-1611788 on Build GRCh38
rs398123612 Chr.6: 1611806-1611807 on Build GRCh38
rs949297601 Chr.6: 1611652-1611652 on Build GRCh38
rs1057519482 Chr.6: 1611710-1611710 on Build GRCh38
rs756037576 Chr.6: 1611781-1611782 on Build GRCh38
rs557912856 Chr.6: 1611783-1611783 on Build GRCh38
rs772035500 Chr.6: 1611691-1611691 on Build GRCh38
rs865901577 Chr.6: 1611656-1611656 on Build GRCh38
rs1011576688 Chr.6: 1611723-1611723 on Build GRCh38
rs794726900 Chr.6: 1611804-1611806 on Build GRCh38
rs759491792 Chr.6: 1611690-1611690 on Build GRCh38
rs1020427428 Chr.6: 1611727-1611727 on Build GRCh38
rs755049427 Chr.6: 1611783-1611788 on Build GRCh38
rs577854700 Chr.6: 1611833-1611833 on Build GRCh38
rs971780796 Chr.6: 1611840-1611842 on Build GRCh38
rs760410240 Chr.6: 1611725-1611725 on Build GRCh38
rs572622655 Chr.6: 1611660-1611660 on Build GRCh38
rs908841297 Chr.6: 1611815-1611815 on Build GRCh38
rs572346201 Chr.6: 1611782-1611783 on Build GRCh38
rs753526474 Chr.6: 1611745-1611745 on Build GRCh38
rs75089353 Chr.6: 1611778-1611778 on Build GRCh38
rs1051933 Chr.6: 1611721-1611721 on Build GRCh38
rs919121619 Chr.6: 1611848-1611848 on Build GRCh38
rs281865460 Chr.6: 1611712-1611712 on Build GRCh38
rs998203463 Chr.6: 1611765-1611765 on Build GRCh38
rs967488377 Chr.6: 1611742-1611742 on Build GRCh38
rs1056318557 Chr.6: 1611698-1611698 on Build GRCh38
rs749942967 Chr.6: 1611850-1611850 on Build GRCh38
rs985824070 Chr.6: 1611804-1611804 on Build GRCh38
rs775432975 Chr.6: 1611720-1611720 on Build GRCh38
rs763885080 Chr.6: 1611744-1611744 on Build GRCh38
rs761435355 Chr.6: 1611764-1611764 on Build GRCh38
rs907221308 Chr.6: 1611683-1611683 on Build GRCh38
rs963143964 Chr.6: 1611825-1611825 on Build GRCh38
rs774351048 Chr.6: 1611657-1611657 on Build GRCh38
rs1001542364 Chr.6: 1611693-1611693 on Build GRCh38
rs886044281 Chr.6: 1611794-1611794 on Build GRCh38

View Full Product Details