Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158578769-158579042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10X1
Entrez Gene ID
128367
Gene Name
olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Gene Aliases
OR1-13, OR1-14, OR10X1P
UniGene
Hs.553589
Gene Chromosome Location
Chr.1: 158578919-158579899 on Build GRCh38
Transcripts
NM_001004477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3476130 c.938C>T p.A313V Chr.1: 158578962-158578962 on Build GRCh38
5634671 c.866T>C p.V289A Chr.1: 158579034-158579034 on Build GRCh38
1269809 c.973A>C p.K325Q Chr.1: 158578927-158578927 on Build GRCh38
3863147 c.911G>A p.S304N Chr.1: 158578989-158578989 on Build GRCh38
5032233 c.887C>T p.P296L Chr.1: 158579013-158579013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748063889 Chr.1: 158579003-158579003 on Build GRCh38
rs1025577711 Chr.1: 158578849-158578849 on Build GRCh38
rs894819530 Chr.1: 158578940-158578940 on Build GRCh38
rs956874468 Chr.1: 158578901-158578901 on Build GRCh38
rs759793557 Chr.1: 158578919-158578919 on Build GRCh38
rs533799212 Chr.1: 158578852-158578852 on Build GRCh38
rs72698782 Chr.1: 158578998-158578998 on Build GRCh38
rs144823498 Chr.1: 158579002-158579002 on Build GRCh38
rs558342760 Chr.1: 158578863-158578863 on Build GRCh38
rs769538674 Chr.1: 158579008-158579008 on Build GRCh38
rs774453422 Chr.1: 158578914-158578914 on Build GRCh38
rs895670830 Chr.1: 158578828-158578828 on Build GRCh38
rs376057452 Chr.1: 158578989-158578989 on Build GRCh38
rs758063971 Chr.1: 158578986-158578986 on Build GRCh38
rs746547371 Chr.1: 158578996-158578996 on Build GRCh38
rs36102561 Chr.1: 158579012-158579012 on Build GRCh38
rs745457271 Chr.1: 158578887-158578887 on Build GRCh38
rs867889881 Chr.1: 158578949-158578949 on Build GRCh38
rs769568708 Chr.1: 158578912-158578912 on Build GRCh38
rs143251974 Chr.1: 158579013-158579013 on Build GRCh38
rs557157625 Chr.1: 158578936-158578936 on Build GRCh38
rs757566670 Chr.1: 158578960-158578960 on Build GRCh38
rs1005671998 Chr.1: 158578877-158578877 on Build GRCh38
rs779525326 Chr.1: 158578888-158578888 on Build GRCh38
rs757589501 Chr.1: 158578871-158578871 on Build GRCh38
rs1047916153 Chr.1: 158578856-158578856 on Build GRCh38
rs1056958953 Chr.1: 158578821-158578821 on Build GRCh38
rs776502941 Chr.1: 158578899-158578899 on Build GRCh38
rs756939060 Chr.1: 158578820-158578820 on Build GRCh38
rs754353122 Chr.1: 158578947-158578947 on Build GRCh38
rs764716071 Chr.1: 158578944-158578944 on Build GRCh38
rs905298187 Chr.1: 158578812-158578812 on Build GRCh38
rs994046633 Chr.1: 158578837-158578837 on Build GRCh38
rs761311226 Chr.1: 158578937-158578937 on Build GRCh38
rs767563329 Chr.1: 158578928-158578928 on Build GRCh38
rs1045152101 Chr.1: 158578791-158578791 on Build GRCh38
rs548128092 Chr.1: 158578810-158578810 on Build GRCh38
rs566435315 Chr.1: 158578833-158578833 on Build GRCh38
rs768560502 Chr.1: 158578891-158578891 on Build GRCh38
rs754269228 Chr.1: 158578858-158578858 on Build GRCh38
rs373159442 Chr.1: 158578965-158578965 on Build GRCh38
rs41273503 Chr.1: 158578890-158578890 on Build GRCh38
rs761628556 Chr.1: 158578909-158578909 on Build GRCh38
rs373728893 Chr.1: 158578854-158578854 on Build GRCh38
rs16840359 Chr.1: 158578882-158578882 on Build GRCh38
rs775440351 Chr.1: 158579012-158579013 on Build GRCh38
rs988345127 Chr.1: 158578941-158578941 on Build GRCh38
rs1018006474 Chr.1: 158578878-158578878 on Build GRCh38
rs750099884 Chr.1: 158578983-158578983 on Build GRCh38

View Full Product Details