Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121420117-121420378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
UniGene
Hs.524800
Gene Chromosome Location
Chr.12: 121402289-121582268 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Gene Symbol
RNF34
Entrez Gene ID
80196
Gene Name
ring finger protein 34
Gene Aliases
CARP-1, CARP1, RFI, RIF, RIFF, hRFI
UniGene
Hs.292804
Gene Chromosome Location
Chr.12: 121400083-121424353 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1224003 c.701A>C p.D234A Chr.12: 121420306-121420306 on Build GRCh38
4039454 c.684C>T p.D228D Chr.12: 121420289-121420289 on Build GRCh38
7313414 c.655G>T p.A219S Chr.12: 121420263-121420263 on Build GRCh38
5588830 c.685G>T p.D229Y Chr.12: 121420290-121420290 on Build GRCh38
936024 c.685G>A p.D229N Chr.12: 121420290-121420290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202132357 Chr.12: 121420286-121420286 on Build GRCh38
rs573048643 Chr.12: 121420274-121420274 on Build GRCh38
rs782340848 Chr.12: 121420287-121420287 on Build GRCh38
rs544697639 Chr.12: 121420322-121420322 on Build GRCh38
rs61739898 Chr.12: 121420310-121420310 on Build GRCh38
rs781802564 Chr.12: 121420326-121420326 on Build GRCh38
rs782676250 Chr.12: 121420333-121420333 on Build GRCh38
rs1036122546 Chr.12: 121420200-121420200 on Build GRCh38
rs558958221 Chr.12: 121420290-121420290 on Build GRCh38
rs138199363 Chr.12: 121420247-121420247 on Build GRCh38
rs147092139 Chr.12: 121420227-121420227 on Build GRCh38
rs782675008 Chr.12: 121420215-121420215 on Build GRCh38
rs782302427 Chr.12: 121420226-121420226 on Build GRCh38
rs111466028 Chr.12: 121420236-121420236 on Build GRCh38
rs782718487 Chr.12: 121420292-121420292 on Build GRCh38
rs536489502 Chr.12: 121420211-121420211 on Build GRCh38
rs782753057 Chr.12: 121420298-121420299 on Build GRCh38
rs782212928 Chr.12: 121420244-121420244 on Build GRCh38
rs782771959 Chr.12: 121420311-121420311 on Build GRCh38
rs782643365 Chr.12: 121420233-121420233 on Build GRCh38
rs782314946 Chr.12: 121420214-121420214 on Build GRCh38
rs782583004 Chr.12: 121420265-121420265 on Build GRCh38
rs1053784242 Chr.12: 121420346-121420346 on Build GRCh38
rs1026196601 Chr.12: 121420254-121420254 on Build GRCh38
rs964455463 Chr.12: 121420302-121420302 on Build GRCh38
rs377017432 Chr.12: 121420289-121420289 on Build GRCh38
rs782167758 Chr.12: 121420298-121420298 on Build GRCh38
rs781893046 Chr.12: 121420295-121420295 on Build GRCh38
rs190921881 Chr.12: 121420283-121420283 on Build GRCh38
rs368005152 Chr.12: 121420323-121420323 on Build GRCh38
rs1001605737 Chr.12: 121420171-121420171 on Build GRCh38
rs905920167 Chr.12: 121420160-121420160 on Build GRCh38
rs992517032 Chr.12: 121420230-121420230 on Build GRCh38
rs782279180 Chr.12: 121420270-121420270 on Build GRCh38
rs61751317 Chr.12: 121420313-121420313 on Build GRCh38
rs574924801 Chr.12: 121420340-121420340 on Build GRCh38
rs782600448 Chr.12: 121420354-121420354 on Build GRCh38

View Full Product Details