Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231197294-231197545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM67
Entrez Gene ID
440730
Gene Name
tripartite motif containing 67
Gene Aliases
TNL
UniGene
Hs.655089
Gene Chromosome Location
Chr.1: 231162928-231221568 on Build GRCh38
Transcripts
NM_001004342.3 NM_001300889.1 XM_011544192.2 XM_017001323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977190 c.1069G>A p.G357R Chr.1: 231197395-231197395 on Build GRCh38
3485071 c.1138C>T p.Q380* Chr.1: 231197464-231197464 on Build GRCh38
4852810 c.1059G>C p.Q353H Chr.1: 231197385-231197385 on Build GRCh38
6808519 c.1105C>A p.L369M Chr.1: 231197431-231197431 on Build GRCh38
1601891 c.1060G>T p.A354S Chr.1: 231197386-231197386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892964182 Chr.1: 231197419-231197419 on Build GRCh38
rs752126953 Chr.1: 231197353-231197353 on Build GRCh38
rs115541663 Chr.1: 231197518-231197518 on Build GRCh38
rs762308140 Chr.1: 231197499-231197500 on Build GRCh38
rs756772976 Chr.1: 231197386-231197386 on Build GRCh38
rs761292553 Chr.1: 231197515-231197515 on Build GRCh38
rs766782845 Chr.1: 231197352-231197352 on Build GRCh38
rs374407015 Chr.1: 231197484-231197484 on Build GRCh38
rs371199106 Chr.1: 231197445-231197445 on Build GRCh38
rs758387844 Chr.1: 231197424-231197424 on Build GRCh38
rs546533927 Chr.1: 231197474-231197474 on Build GRCh38
rs773470834 Chr.1: 231197333-231197333 on Build GRCh38
rs779763083 Chr.1: 231197441-231197441 on Build GRCh38
rs776054889 Chr.1: 231197488-231197488 on Build GRCh38
rs759951398 Chr.1: 231197487-231197487 on Build GRCh38
rs866850596 Chr.1: 231197426-231197426 on Build GRCh38
rs781075562 Chr.1: 231197463-231197463 on Build GRCh38
rs753331551 Chr.1: 231197382-231197382 on Build GRCh38
rs553825799 Chr.1: 231197428-231197428 on Build GRCh38
rs374943785 Chr.1: 231197449-231197449 on Build GRCh38
rs950698296 Chr.1: 231197406-231197406 on Build GRCh38
rs769914905 Chr.1: 231197478-231197478 on Build GRCh38
rs368412114 Chr.1: 231197486-231197486 on Build GRCh38
rs758189294 Chr.1: 231197433-231197433 on Build GRCh38
rs773312756 Chr.1: 231197483-231197483 on Build GRCh38
rs763732446 Chr.1: 231197369-231197369 on Build GRCh38
rs1036138547 Chr.1: 231197376-231197376 on Build GRCh38
rs760074407 Chr.1: 231197356-231197356 on Build GRCh38
rs376886233 Chr.1: 231197415-231197415 on Build GRCh38
rs763295690 Chr.1: 231197334-231197334 on Build GRCh38

View Full Product Details