Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37867771-37867996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5794192 c.707C>T p.P236L Chr.7: 37867787-37867787 on Build GRCh38
3229114 c.752G>A p.R251Q Chr.7: 37867832-37867832 on Build GRCh38
3229113 c.725C>A p.P242Q Chr.7: 37867805-37867805 on Build GRCh38
7331874 c.813A>G p.Q271Q Chr.7: 37867893-37867893 on Build GRCh38
3638521 c.774C>T p.V258V Chr.7: 37867854-37867854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371745920 Chr.7: 37867827-37867827 on Build GRCh38
rs1044473414 Chr.7: 37867855-37867855 on Build GRCh38
rs542596382 Chr.7: 37867964-37867964 on Build GRCh38
rs975118756 Chr.7: 37867815-37867815 on Build GRCh38
rs183829557 Chr.7: 37867955-37867955 on Build GRCh38
rs545236479 Chr.7: 37867938-37867938 on Build GRCh38
rs139516225 Chr.7: 37867819-37867819 on Build GRCh38
rs757897300 Chr.7: 37867804-37867804 on Build GRCh38
rs768715665 Chr.7: 37867821-37867821 on Build GRCh38
rs140957789 Chr.7: 37867803-37867803 on Build GRCh38
rs918829851 Chr.7: 37867892-37867892 on Build GRCh38
rs965048909 Chr.7: 37867813-37867813 on Build GRCh38
rs868314762 Chr.7: 37867861-37867861 on Build GRCh38
rs201277639 Chr.7: 37867822-37867822 on Build GRCh38
rs116545712 Chr.7: 37867925-37867925 on Build GRCh38
rs780346927 Chr.7: 37867862-37867862 on Build GRCh38
rs779108426 Chr.7: 37867866-37867866 on Build GRCh38
rs773181802 Chr.7: 37867934-37867934 on Build GRCh38
rs759117674 Chr.7: 37867899-37867900 on Build GRCh38
rs118137117 Chr.7: 37867954-37867954 on Build GRCh38
rs760474274 Chr.7: 37867830-37867830 on Build GRCh38
rs781400657 Chr.7: 37867847-37867847 on Build GRCh38
rs369365068 Chr.7: 37867811-37867811 on Build GRCh38
rs764505957 Chr.7: 37867854-37867854 on Build GRCh38
rs572433461 Chr.7: 37867959-37867959 on Build GRCh38
rs749510266 Chr.7: 37867863-37867863 on Build GRCh38
rs948721664 Chr.7: 37867840-37867840 on Build GRCh38
rs776246131 Chr.7: 37867881-37867883 on Build GRCh38
rs200131495 Chr.7: 37867833-37867833 on Build GRCh38
rs755131627 Chr.7: 37867864-37867864 on Build GRCh38
rs199973571 Chr.7: 37867823-37867823 on Build GRCh38
rs778607368 Chr.7: 37867801-37867801 on Build GRCh38
rs773058604 Chr.7: 37867828-37867828 on Build GRCh38
rs375200127 Chr.7: 37867800-37867800 on Build GRCh38
rs866906346 Chr.7: 37867868-37867868 on Build GRCh38
rs748454007 Chr.7: 37867848-37867848 on Build GRCh38
rs201987014 Chr.7: 37867832-37867832 on Build GRCh38
rs553733754 Chr.7: 37867852-37867852 on Build GRCh38
rs746203531 Chr.7: 37867809-37867809 on Build GRCh38
rs746962929 Chr.7: 37867939-37867939 on Build GRCh38
rs904617359 Chr.7: 37867895-37867895 on Build GRCh38
rs764828685 Chr.7: 37867845-37867845 on Build GRCh38
rs371341539 Chr.7: 37867891-37867891 on Build GRCh38
rs113330102 Chr.7: 37867831-37867831 on Build GRCh38
rs201321298 Chr.7: 37867818-37867818 on Build GRCh38
rs781768255 Chr.7: 37867807-37867807 on Build GRCh38
rs895815053 Chr.7: 37867839-37867839 on Build GRCh38
rs191578224 Chr.7: 37867873-37867873 on Build GRCh38
rs867863283 Chr.7: 37867843-37867843 on Build GRCh38
rs756469390 Chr.7: 37867856-37867856 on Build GRCh38

View Full Product Details