Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5635117-5635384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
UniGene
Hs.148970
Gene Chromosome Location
Chr.12: 5560241-5948790 on Build GRCh38
Transcripts
NM_001278596.1 NM_001278597.1 XM_006718988.3 XM_011520975.2 XM_011520978.2 XM_017019672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749223 c.1667G>A p.R556Q Chr.12: 5635304-5635304 on Build GRCh38
7400146 c.1752C>T p.I584I Chr.12: 5635219-5635219 on Build GRCh38
7400149 c.1749C>T p.I583I Chr.12: 5635219-5635219 on Build GRCh38
6398814 c.1798G>T p.A600S Chr.12: 5635173-5635173 on Build GRCh38
3463641 c.1787G>A p.G596D Chr.12: 5635181-5635181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368385506 Chr.12: 5635327-5635327 on Build GRCh38
rs756083891 Chr.12: 5635263-5635263 on Build GRCh38
rs893053349 Chr.12: 5635353-5635353 on Build GRCh38
rs779424459 Chr.12: 5635287-5635287 on Build GRCh38
rs374184483 Chr.12: 5635311-5635311 on Build GRCh38
rs747406275 Chr.12: 5635248-5635248 on Build GRCh38
rs755452999 Chr.12: 5635253-5635253 on Build GRCh38
rs750512780 Chr.12: 5635235-5635235 on Build GRCh38
rs750456230 Chr.12: 5635139-5635139 on Build GRCh38
rs762198146 Chr.12: 5635138-5635138 on Build GRCh38
rs900609306 Chr.12: 5635328-5635328 on Build GRCh38
rs886446638 Chr.12: 5635309-5635309 on Build GRCh38
rs763415543 Chr.12: 5635307-5635307 on Build GRCh38
rs754864698 Chr.12: 5635168-5635168 on Build GRCh38
rs200078432 Chr.12: 5635329-5635329 on Build GRCh38
rs765593244 Chr.12: 5635232-5635232 on Build GRCh38
rs755921891 Chr.12: 5635340-5635340 on Build GRCh38
rs373762182 Chr.12: 5635292-5635292 on Build GRCh38
rs373059086 Chr.12: 5635180-5635180 on Build GRCh38
rs777455201 Chr.12: 5635182-5635182 on Build GRCh38
rs773771654 Chr.12: 5635304-5635304 on Build GRCh38
rs539422592 Chr.12: 5635150-5635150 on Build GRCh38
rs768954741 Chr.12: 5635279-5635282 on Build GRCh38
rs752551509 Chr.12: 5635333-5635333 on Build GRCh38
rs368489336 Chr.12: 5635299-5635299 on Build GRCh38
rs777147988 Chr.12: 5635349-5635352 on Build GRCh38
rs749888230 Chr.12: 5635230-5635230 on Build GRCh38
rs199769145 Chr.12: 5635358-5635358 on Build GRCh38
rs769108613 Chr.12: 5635212-5635212 on Build GRCh38
rs766437380 Chr.12: 5635250-5635250 on Build GRCh38
rs781061139 Chr.12: 5635262-5635262 on Build GRCh38
rs201631550 Chr.12: 5635284-5635284 on Build GRCh38
rs759111246 Chr.12: 5635197-5635197 on Build GRCh38
rs776841463 Chr.12: 5635213-5635213 on Build GRCh38
rs749224901 Chr.12: 5635274-5635274 on Build GRCh38
rs112073363 Chr.12: 5635169-5635169 on Build GRCh38
rs372864505 Chr.12: 5635305-5635305 on Build GRCh38
rs971133766 Chr.12: 5635223-5635223 on Build GRCh38
rs370829559 Chr.12: 5635251-5635251 on Build GRCh38
rs377359247 Chr.12: 5635183-5635183 on Build GRCh38
rs191188083 Chr.12: 5635217-5635217 on Build GRCh38
rs748165730 Chr.12: 5635179-5635179 on Build GRCh38
rs376269086 Chr.12: 5635192-5635192 on Build GRCh38
rs1047719982 Chr.12: 5635306-5635306 on Build GRCh38
rs77241305 Chr.12: 5635355-5635355 on Build GRCh38
rs374634976 Chr.12: 5635242-5635242 on Build GRCh38
rs370127571 Chr.12: 5635288-5635288 on Build GRCh38
rs770632937 Chr.12: 5635280-5635280 on Build GRCh38
rs201079863 Chr.12: 5635189-5635189 on Build GRCh38
rs755132323 Chr.12: 5635256-5635256 on Build GRCh38
rs748401795 Chr.12: 5635295-5635295 on Build GRCh38
rs757241441 Chr.12: 5635360-5635360 on Build GRCh38
rs751764238 Chr.12: 5635162-5635162 on Build GRCh38
rs944779396 Chr.12: 5635310-5635310 on Build GRCh38
rs774365666 Chr.12: 5635326-5635326 on Build GRCh38
rs780189692 Chr.12: 5635161-5635161 on Build GRCh38

View Full Product Details