Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1525087-1525318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3A
Entrez Gene ID
55210
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
-
UniGene
Hs.23413
Gene Chromosome Location
Chr.1: 1512143-1534687 on Build GRCh38
Transcripts
NM_001170535.1 NM_001170536.1 NM_018188.3 XM_017001628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606394 c.1215G>A p.K405K Chr.1: 1525284-1525284 on Build GRCh38
6606393 c.1403G>A p.R468Q Chr.1: 1525284-1525284 on Build GRCh38
5156689 c.1359-6T>C p.? Chr.1: 1525234-1525234 on Build GRCh38
1245624 c.1400A>T p.K467M Chr.1: 1525281-1525281 on Build GRCh38
5934225 c.1409C>T p.T470I Chr.1: 1525290-1525290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368010141 Chr.1: 1525171-1525171 on Build GRCh38
rs11260607 Chr.1: 1525217-1525217 on Build GRCh38
rs544185941 Chr.1: 1525161-1525161 on Build GRCh38
rs564903814 Chr.1: 1525137-1525137 on Build GRCh38
rs756351187 Chr.1: 1525227-1525227 on Build GRCh38
rs371591033 Chr.1: 1525176-1525176 on Build GRCh38
rs979842020 Chr.1: 1525132-1525132 on Build GRCh38
rs112869160 Chr.1: 1525234-1525234 on Build GRCh38
rs138094382 Chr.1: 1525264-1525264 on Build GRCh38
rs765378350 Chr.1: 1525288-1525288 on Build GRCh38
rs10465902 Chr.1: 1525201-1525201 on Build GRCh38
rs764521406 Chr.1: 1525212-1525212 on Build GRCh38
rs757205195 Chr.1: 1525292-1525292 on Build GRCh38
rs188675569 Chr.1: 1525229-1525229 on Build GRCh38
rs889401370 Chr.1: 1525169-1525169 on Build GRCh38
rs149995099 Chr.1: 1525291-1525291 on Build GRCh38
rs746922606 Chr.1: 1525232-1525232 on Build GRCh38
rs968111601 Chr.1: 1525118-1525118 on Build GRCh38
rs762348060 Chr.1: 1525268-1525268 on Build GRCh38
rs561188633 Chr.1: 1525165-1525165 on Build GRCh38
rs767831894 Chr.1: 1525269-1525269 on Build GRCh38
rs372683595 Chr.1: 1525170-1525170 on Build GRCh38
rs11260606 Chr.1: 1525197-1525197 on Build GRCh38
rs116637550 Chr.1: 1525210-1525210 on Build GRCh38
rs374564620 Chr.1: 1525195-1525195 on Build GRCh38
rs376544187 Chr.1: 1525179-1525179 on Build GRCh38
rs545112242 Chr.1: 1525114-1525114 on Build GRCh38
rs554255312 Chr.1: 1525263-1525263 on Build GRCh38
rs544239006 Chr.1: 1525253-1525253 on Build GRCh38
rs1049184470 Chr.1: 1525166-1525166 on Build GRCh38
rs201906120 Chr.1: 1525214-1525214 on Build GRCh38
rs888796743 Chr.1: 1525235-1525235 on Build GRCh38
rs199697769 Chr.1: 1525215-1525215 on Build GRCh38
rs112376265 Chr.1: 1525144-1525144 on Build GRCh38
rs902029914 Chr.1: 1525180-1525180 on Build GRCh38
rs57871049 Chr.1: 1525154-1525154 on Build GRCh38
rs559898799 Chr.1: 1525172-1525172 on Build GRCh38
rs386627521 Chr.1: 1525201-1525202 on Build GRCh38
rs1021037373 Chr.1: 1525185-1525185 on Build GRCh38
rs376153971 Chr.1: 1525283-1525283 on Build GRCh38
rs991393162 Chr.1: 1525138-1525138 on Build GRCh38
rs201199935 Chr.1: 1525211-1525211 on Build GRCh38
rs532495854 Chr.1: 1525182-1525182 on Build GRCh38
rs367956062 Chr.1: 1525189-1525189 on Build GRCh38
rs139254288 Chr.1: 1525277-1525277 on Build GRCh38
rs781036576 Chr.1: 1525249-1525249 on Build GRCh38
rs201577129 Chr.1: 1525230-1525230 on Build GRCh38
rs949825610 Chr.1: 1525152-1525152 on Build GRCh38
rs760819756 Chr.1: 1525278-1525278 on Build GRCh38
rs759197355 Chr.1: 1525208-1525208 on Build GRCh38
rs925918338 Chr.1: 1525134-1525134 on Build GRCh38
rs559935599 Chr.1: 1525284-1525284 on Build GRCh38
rs917106809 Chr.1: 1525149-1525149 on Build GRCh38
rs111328913 Chr.1: 1525226-1525226 on Build GRCh38
rs10465906 Chr.1: 1525202-1525202 on Build GRCh38
rs375954457 Chr.1: 1525231-1525231 on Build GRCh38
rs373058267 Chr.1: 1525267-1525267 on Build GRCh38
rs575132198 Chr.1: 1525252-1525252 on Build GRCh38
rs764360847 Chr.1: 1525126-1525126 on Build GRCh38
rs550766381 Chr.1: 1525164-1525164 on Build GRCh38

View Full Product Details