Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24823144-24823417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019189 c.2862A>T p.E954D Chr.2: 24823230-24823230 on Build GRCh38
6410561 c.2775G>T p.M925I Chr.2: 24823317-24823317 on Build GRCh38
1182022 c.2840A>T p.E947V Chr.2: 24823252-24823252 on Build GRCh38
5966405 c.2839G>T p.E947* Chr.2: 24823253-24823253 on Build GRCh38
3839147 c.2749C>A p.Q917K Chr.2: 24823343-24823343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746758858 Chr.2: 24823243-24823243 on Build GRCh38
rs764845615 Chr.2: 24823362-24823362 on Build GRCh38
rs541099242 Chr.2: 24823197-24823197 on Build GRCh38
rs758331959 Chr.2: 24823240-24823240 on Build GRCh38
rs751552916 Chr.2: 24823372-24823372 on Build GRCh38
rs762266451 Chr.2: 24823324-24823324 on Build GRCh38
rs767970710 Chr.2: 24823170-24823170 on Build GRCh38
rs760980598 Chr.2: 24823335-24823335 on Build GRCh38
rs549761443 Chr.2: 24823380-24823380 on Build GRCh38
rs756699487 Chr.2: 24823222-24823222 on Build GRCh38
rs771128870 Chr.2: 24823272-24823272 on Build GRCh38
rs1127568 Chr.2: 24823221-24823221 on Build GRCh38
rs764798957 Chr.2: 24823224-24823224 on Build GRCh38
rs143693626 Chr.2: 24823338-24823338 on Build GRCh38
rs887841958 Chr.2: 24823227-24823227 on Build GRCh38
rs367903449 Chr.2: 24823263-24823263 on Build GRCh38
rs772602552 Chr.2: 24823293-24823293 on Build GRCh38
rs889563269 Chr.2: 24823358-24823359 on Build GRCh38
rs776200284 Chr.2: 24823339-24823339 on Build GRCh38
rs757149353 Chr.2: 24823387-24823387 on Build GRCh38
rs368676965 Chr.2: 24823369-24823369 on Build GRCh38
rs760088030 Chr.2: 24823169-24823169 on Build GRCh38
rs754749699 Chr.2: 24823376-24823376 on Build GRCh38
rs761512893 Chr.2: 24823351-24823351 on Build GRCh38
rs757482992 Chr.2: 24823386-24823386 on Build GRCh38
rs199667987 Chr.2: 24823370-24823370 on Build GRCh38
rs763699009 Chr.2: 24823198-24823198 on Build GRCh38
rs754839463 Chr.2: 24823254-24823254 on Build GRCh38
rs745923661 Chr.2: 24823393-24823393 on Build GRCh38
rs34134504 Chr.2: 24823309-24823310 on Build GRCh38
rs772069036 Chr.2: 24823394-24823394 on Build GRCh38
rs779895158 Chr.2: 24823241-24823241 on Build GRCh38
rs932158367 Chr.2: 24823325-24823325 on Build GRCh38
rs776009447 Chr.2: 24823311-24823311 on Build GRCh38
rs959109538 Chr.2: 24823389-24823389 on Build GRCh38
rs763339528 Chr.2: 24823260-24823260 on Build GRCh38
rs375101795 Chr.2: 24823290-24823290 on Build GRCh38
rs774480624 Chr.2: 24823282-24823282 on Build GRCh38
rs749959260 Chr.2: 24823230-24823230 on Build GRCh38
rs897000384 Chr.2: 24823194-24823194 on Build GRCh38

View Full Product Details