Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50323767-50324028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC3
Entrez Gene ID
3748
Gene Name
potassium voltage-gated channel subfamily C member 3
Gene Aliases
KSHIIID, KV3.3, SCA13
UniGene
Hs.467146
Gene Chromosome Location
Chr.19: 50311942-50333515 on Build GRCh38
Transcripts
NM_004977.2 NR_110912.1 XM_006723203.2 XM_011526925.2 XM_011526926.1 XM_011526928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6560802 c.1092C>A p.L364L Chr.19: 50323861-50323861 on Build GRCh38
6560803 c.1011C>G p.I337M Chr.19: 50323942-50323942 on Build GRCh38
4694227 c.1135_1136insA p.S379fs*92 Chr.19: 50323817-50323818 on Build GRCh38
5810049 c.1006A>G p.N336D Chr.19: 50323947-50323947 on Build GRCh38
4779786 c.991G>A p.G331R Chr.19: 50323962-50323962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781277310 Chr.19: 50323945-50323945 on Build GRCh38
rs145167102 Chr.19: 50323951-50323951 on Build GRCh38
rs755008922 Chr.19: 50323940-50323940 on Build GRCh38
rs368239113 Chr.19: 50323893-50323893 on Build GRCh38
rs115351343 Chr.19: 50323935-50323935 on Build GRCh38
rs376287269 Chr.19: 50323843-50323843 on Build GRCh38
rs114192110 Chr.19: 50323969-50323969 on Build GRCh38
rs758088145 Chr.19: 50323922-50323922 on Build GRCh38
rs115543544 Chr.19: 50323801-50323801 on Build GRCh38
rs1011930554 Chr.19: 50323849-50323849 on Build GRCh38
rs746999292 Chr.19: 50323816-50323816 on Build GRCh38
rs866174515 Chr.19: 50324004-50324004 on Build GRCh38
rs146613004 Chr.19: 50323902-50323902 on Build GRCh38
rs571327522 Chr.19: 50323921-50323921 on Build GRCh38
rs767951843 Chr.19: 50323882-50323882 on Build GRCh38
rs141390395 Chr.19: 50323911-50323911 on Build GRCh38
rs761084175 Chr.19: 50323987-50323987 on Build GRCh38
rs879020085 Chr.19: 50323861-50323861 on Build GRCh38
rs146772778 Chr.19: 50323963-50323963 on Build GRCh38
rs150426801 Chr.19: 50324002-50324002 on Build GRCh38
rs199908446 Chr.19: 50323887-50323887 on Build GRCh38
rs757536869 Chr.19: 50323794-50323794 on Build GRCh38
rs201662217 Chr.19: 50323903-50323903 on Build GRCh38
rs776765452 Chr.19: 50323879-50323879 on Build GRCh38
rs970287045 Chr.19: 50323889-50323889 on Build GRCh38
rs749005904 Chr.19: 50323967-50323967 on Build GRCh38
rs200905864 Chr.19: 50323970-50323970 on Build GRCh38
rs142937043 Chr.19: 50323873-50323873 on Build GRCh38
rs144968007 Chr.19: 50323789-50323789 on Build GRCh38
rs1029753390 Chr.19: 50323944-50323944 on Build GRCh38
rs769502387 Chr.19: 50323856-50323856 on Build GRCh38
rs768028091 Chr.19: 50323830-50323830 on Build GRCh38
rs772956266 Chr.19: 50323965-50323965 on Build GRCh38
rs1000435321 Chr.19: 50323915-50323915 on Build GRCh38
rs961422558 Chr.19: 50323918-50323918 on Build GRCh38
rs773009246 Chr.19: 50323870-50323870 on Build GRCh38
rs761550680 Chr.19: 50323846-50323846 on Build GRCh38
rs868826741 Chr.19: 50323960-50323960 on Build GRCh38
rs771916263 Chr.19: 50323806-50323806 on Build GRCh38
rs763237050 Chr.19: 50324005-50324005 on Build GRCh38
rs778679027 Chr.19: 50323800-50323800 on Build GRCh38
rs761297836 Chr.19: 50323888-50323888 on Build GRCh38
rs200893614 Chr.19: 50323807-50323807 on Build GRCh38
rs371799397 Chr.19: 50323936-50323936 on Build GRCh38

View Full Product Details